Merck
  • Synthesis, Photophysical and Redox Properties of the D-π-A Type Pyrimidine Dyes Bearing the 9-Phenyl-9H-Carbazole Moiety.
Synthesis, Photophysical and Redox Properties of the D-π-A Type Pyrimidine Dyes Bearing the 9-Phenyl-9H-Carbazole Moiety.
Journal of fluorescence (2015-03-31)
Egor V Verbitskiy, Aleksandr V Schepochkin, Nadezhda I Makarova, Igor V Dorogan, Anatoly V Metelitsa, Vladimir I Minkin, Sergey A Kozyukhin, Victor V Emets, Vitaly A Grindberg, Oleg N Chupakhin, Gennady L Rusinov, Valery N Charushin
ZUSAMMENFASSUNG
Novel donor-π-acceptor dyes bearing the pyrimidine unit as an electron-withdrawing group have been synthesized by using combination of two processes, based on the microwave-assisted Suzuki cross-coupling reaction and nucleophilic aromatic substitution of hydrogen. Spectral properties of the obtained dyes in six aprotic solvents of various polarities have been studied by ultraviolet-visible and fluorescence spectroscopy. In contrast to the absorption spectra, fluorescence emission spectra displayed a strong dependence from their solvent polarities. The nature of the observed long wavelength maxima has been elucidated by means of quantum chemical calculations. The electrochemical properties of these dyes have been investigated by using cyclic voltammetry, while their photovoltaic performance was evaluated by a device fabrication study. The experimental and calculation data show that all of the dyes can be regarded as potentially good photosensitizers for dye-sensitized solar cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Pinakon, 98%
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
Argon, ≥99.998%
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O
Sigma-Aldrich
Argon-40Ar, 99.95 atom %