Merck
  • Development of fluorescent peptide substrates and assays for the key autophagy-initiating cysteine protease enzyme, ATG4B.
Development of fluorescent peptide substrates and assays for the key autophagy-initiating cysteine protease enzyme, ATG4B.
Bioorganic & medicinal chemistry (2015-05-17)
Lubomir Vezenkov, Nicolette S Honson, Nag S Kumar, Damien Bosc, Suzana Kovacic, Thanh G Nguyen, Tom A Pfeifer, Robert N Young
ZUSAMMENFASSUNG
An efficient assay for monitoring the activity of the key autophagy-initiating enzyme ATG4B based on a small peptide substrate has been developed. A number of putative small fluorogenic peptide substrates were prepared and evaluated and optimized compounds showed reasonable rates of cleavage but required high enzyme concentrations which limited their value. A modified peptide substrate incorporating a less sterically demanding self-immolative element was designed and synthesized and was shown to have enhanced properties useful for evaluating inhibitors of ATG4B. Substrate cleavage was readily monitored and was linear for up to 4h but enzyme concentrations of about ten-fold higher were required compared to assays using protein substrate LC3 or analogs thereof (such as FRET-LC3). Several known inhibitors of ATG4B were evaluated using the small peptide substrate and gave IC50 values 3-7 fold higher than previously obtained values using the FRET-LC3 substrate.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Acetylchlorid, reagent grade, 98%
Sigma-Aldrich
Acetylchlorid, reagent grade, 98%
Sigma-Aldrich
Aurintri-Carboxylsäure, practical grade, ≥85% (titration), powder
Sigma-Aldrich
Hypericin aus Hypericum perforatum, ~95% (HPLC)
Sigma-Aldrich
Acetylchlorid -Lösung, 1 M in methylene chloride
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®