Merck
  • Determination of perfluorinated sulfonate and perfluorinated acids in tissues of free-living European beaver (castor fiber L.) by d-SPE/ micro-UHPLC-MS/MS.
Determination of perfluorinated sulfonate and perfluorinated acids in tissues of free-living European beaver (castor fiber L.) by d-SPE/ micro-UHPLC-MS/MS.
Ecotoxicology and environmental safety (2015-07-06)
Magdalena Surma, Zygmunt Giżejewski, Henryk Zieliński
ZUSAMMENFASSUNG
Perfluorooctanesulfonate (PFOS) and perfluorooctanoic acid (PFOA) are the main representatives of an rising class of persistent organic pollutants (POPs), perfluorochemicals (PFCs). In this study, determination of selected PFCs concentration in liver, brain, tail, adipose and peritoneum tissues of free-living European beaver (Castor fiber L.) was addressed. Tissue samples, collected from beavers living in Masurian Lakeland (NE Poland), were analyzed by dispersive Solid Phase Extraction (d-SPE) with micro-UHPLC-MS/MS system. In a group of ten selected pefrluorinated compounds only two perfluorinated acids (PFOA and PFNA) and one perfluorinated sulfonate (PFOS) were quantified. PFOA was detected in all analysed tissue samples in both female and male beavers in a range from 0.55 to 0.98ngg(-1) ww whereas PFOS was identified in all analyzed female beaver tissues and only in liver, subcutaneous adipose and peritoneum tissues of male beavers at the concentration level from 0.86 to 5.08ngg(-1) ww. PFNA was only identified in female beaver tissues (liver, subcutaneous adipose and peritoneum) in a range from 1.50 to 6.61ngg(-1) ww. This study demonstrated the bioaccumulation of PFCs in tissue samples collected from beavers living in area known as green lungs of Poland. The results provided in this study indicate for the increasing risk of PFCs occurrence in the environment and the level of PFCs in tissue of free-living European beavers may serve as bioindicator of environmental pollution by these compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Perfluoroctansäure, 95%
Sigma-Aldrich
Divinylbenzol, technical grade, 55%
Sigma-Aldrich
Divinylbenzol, technical grade, 80%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
Kohlenstoff, mesoporös, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Octadecylsilan, 97%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
Methan-12C, 99.99 atom % 12C
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon