Merck
  • MiR-31 modulates coelomocytes ROS production via targeting p105 in Vibrio splendidus challenged sea cucumber Apostichopus japonicus in vitro and in vivo.
MiR-31 modulates coelomocytes ROS production via targeting p105 in Vibrio splendidus challenged sea cucumber Apostichopus japonicus in vitro and in vivo.
Fish & shellfish immunology (2015-04-29)
Meng Lu, Pengjuan Zhang, Chenghua Li, Weiwei Zhang, Chunhua Jin, Qingxi Han
ZUSAMMENFASSUNG
MiR-31 is a critical regulator of gene expression in many pathogenic processes in vertebrates. In this study, we identified p105 as a novel target of miR-31 in Apostichopus japonicus and investigated their regulatory roles in vitro and in vivo. The negative expression profiles between miR-31 and Ajp105 were detected in both LPS-exposed primary coelomocytes and Vibrio splendidus-challenged sea cucumber. Co-infection miR-31 mimics significantly depressed the expression of Ajp105 and increased ROS production in vitro. In contrast, miR-31 inhibitor significantly elevated the expression of Ajp105 and decreased ROS level. Consistently, miR-31 over-expression or Ajp105 silencing in vivo both greatly promoted ROS accumulation. Taken together, our findings confirmed that miR-31 could modulate respiratory burst via targeting Ajp105 during sea cucumber pathological development.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder
Sigma-Aldrich
Streptomycin -sulfat (Salz), powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Streptomycin, Ready Made Solution, 100 mg/mL in water