Merck
  • Targeted multidrug-resistance reversal in tumor based on PEG-PLL-PLGA polymer nano drug delivery system.
Targeted multidrug-resistance reversal in tumor based on PEG-PLL-PLGA polymer nano drug delivery system.
International journal of nanomedicine (2015-07-28)
Liting Guo, Haijun Zhang, Fei Wang, Ping Liu, Yonglu Wang, Guohua Xia, Ran Liu, Xueming Li, Haixiang Yin, Hulin Jiang, Baoan Chen
ZUSAMMENFASSUNG
The study investigated the reversal of multidrug resistance (MDR) and the biodistribution of nanoparticles (NPs) that target leukemia cells in a nude mice model via a surface-bound transferrin (Tf). The cytotoxic cargo of daunorubicin (DNR) and tetrandrine (Tet) was protected in the NPs by an outer coat composed of polyethylene glycol (PEG)-poly-L-lysine (PLL)-poly(lactic-co-glycolic acid) (PLGA) NPs. Injection of DNR-Tet-Tf-PEG-PLL-PLGA NPs into nude mice bearing MDR leukemia cell K562/A02 xenografts was shown to inhibit tumor growth, and contemporaneous immunohistochemical analysis of tumor tissue showed the targeted NPs induced apoptosis in tumor cells. Targeted tumor cells exhibited a marked increase in Tf receptor expression, with noticeable decreases in P-glycoprotein, MDR protein, and nuclear factor κB, as assessed by quantitative real-time polymerase chain reaction and Western blot analysis. Moreover, the concentration of DNR was shown to increase in plasma, tumor tissue, and major organs. Flow cytometry analysis with a near-infrared fluorescent (NIRF) dye, NIR797, was used to study the effectiveness of Tf as a targeting group for leukemia cells, a finding that was supported by NIRF imaging in tumor-bearing nude mice. In summary, our studies show that DNR-Tet-Tf-PEG-PLL-PLGA NPs provide a specific and effective means to target cytotoxic drugs to MDR tumor cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
4-(Dimethylamino)-pyridin, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid, 99%
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Milchsäure, meets USP testing specifications
Sigma-Aldrich
Glycolsäure, ReagentPlus®, 99%
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
1,1′-Carbonyldiimidazol, reagent grade
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
1,1′-Carbonyldiimidazol, ≥97.0% (T)
Sigma-Aldrich
N,N′-Dicyclohexylcarbodiimid -Lösung, 1.0 M in methylene chloride
Sigma-Aldrich
Glycolsäure -Lösung, technical grade, 70 wt. % in H2O
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup
Sigma-Aldrich
Glycolsäure -Lösung, high purity, 70 wt. % in H2O
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-Glycerin, ≥95% (TLC), liquid
Sigma-Aldrich
Glycolsäure, BioXtra, ≥98.0% (titration)