Merck
  • Speciation of inorganic arsenic species and total inorganic arsenic in rice using microwave-assisted dispersive liquid-liquid micro-extraction and electrothermal atomic absorption spectrometry.
Speciation of inorganic arsenic species and total inorganic arsenic in rice using microwave-assisted dispersive liquid-liquid micro-extraction and electrothermal atomic absorption spectrometry.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment (2015-05-12)
Toraj Ahmadi-Jouibari, Nazir Fattahi
ZUSAMMENFASSUNG
Human exposure to inorganic arsenic (iAs) via rice consumption is of increasing concern. In the present study, microwave-assisted dispersive liquid-liquid micro-extraction (MADLLME) and electrothermal atomic absorption spectrometry (ETAAS) were developed for the speciation of iAs in rice samples. After microwave-assisted digestion, the As(III) ion reacted with diethyldithiophosphoric acid (DDTP) to form an As-DDTP complex and was extracted at the same time. Some parameters affecting digestion, complex formation, and extraction were studied and optimised. Under the optimised conditions, a detection limit of 0.2 µg kg(-1) with a correlation coefficient of 0.9901 were obtained with a calibration curve in the range of 0.5-200 µg kg(-1). Total iAs was determined after reduction of As(V) to As(III) with sodium thiosulfate and potassium iodide, and As(V) was calculated by difference. The proposed extraction procedure was successfully applied for the determination of iAs ions in certified reference materials (NIST CRM 1568a and NMIJ CRM 7503a) and 10 rice samples produced in Iran and other Asian countries.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salpetersäure, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Kaliumiodid, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumthiosulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Salpetersäure, ACS reagent, ≥90.0%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Kaliumiodid, ≥99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumthiosulfat -Lösung, 2 g/dL in deionized water