Merck
  • Redox-active biflavonoids from Garcinia brasiliensis as inhibitors of neutrophil oxidative burst and human erythrocyte membrane damage.
Redox-active biflavonoids from Garcinia brasiliensis as inhibitors of neutrophil oxidative burst and human erythrocyte membrane damage.
Journal of ethnopharmacology (2015-09-01)
Phanuel Saroni Arwa, Maria Luiza Zeraik, Valdecir Farias Ximenes, Luiz Marcos da Fonseca, Vanderlan da Silva Bolzani, Dulce Helena Siqueira Silva
ZUSAMMENFASSUNG
Garcinia brasiliensis, a plant native to the Brazilian Amazon Rainforest, is used in traditional medicine to treat inflammation of the urinary tract, peptic ulcers, arthritis and other conditions. The purposes of this study were to analyze the chemical constituents of G. brasiliensis branches and leaves and to evaluate the potential of isolated compounds to act as inhibitors of both the oxidative burst of stimulated neutrophils and oxidative damage in human erythrocyte membranes to verify the antioxidant and anti-inflammatory effects of this plant. Neutrophils were isolated from the blood of healthy donors by Ficoll-Paque density gradient centrifugation. Superoxide anion and total reactive oxygen species (ROS) produced by stimulated neutrophils were measured by WST-1 reduction and luminol-enhanced chemiluminescence assays, respectively. Radical-induced lipoperoxidation and hemolysis were performed using erythrocytes from the blood of healthy donors. Compounds were isolated from G. brasiliensis branches and leaves by HPLC microfractionation, and structure elucidation of the isolated compounds was performed based on NMR and HR-MS analyses. The biflavonoids procyanidin, fukugetin, amentoflavone and podocarpusflavone isolated from G. brasiliensis showed potent inhibitory effects on the oxidative burst of human neutrophils, inhibiting ROS production by 50% at 1 μmol L(-1). These biflavonoids also proved to be potent inhibitors of hemolysis (with 88 ± 7% inhibition at 50 µmol L(-1) for procyanidin) and lipid peroxidation in human erythrocytes, with a malondialdehyde level (a biomarker of oxidative stress) of 8.5 ± 0.3 nmol/mg Hb at 50 µmol L(-1) for procyanidin. These findings indicate that the biflavonoids extracted from G. brasiliensis branches and leaves modulate oxidative stress via inhibition of NADPH oxidase and ROS production by stimulated human neutrophils. Furthermore, the biflavonoids exhibited potent inhibition of oxidant hemolysis and lipid peroxidation induced by AAPH in human erythrocytes. Therefore, these studies suggest the use of G. brasiliensis extract as an antioxidant and anti-inflammatory agent.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylacetat, ACS reagent, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
2,2-Diphenyl-1-Picrylhydrazyl
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Trichloressigsäure, ACS reagent, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Trichloressigsäure -Lösung, 6.1 N
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥99%
Sigma-Aldrich
Luminol, 97%
Sigma-Aldrich
2-Thiobarbitursäure, ≥98%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Trichloressigsäure, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Luminol, ≥97% (HPLC)
Sigma-Aldrich
3,3′,5,5′-Tetramethylbenzidin, ≥98% (TLC)
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent