Merck
  • Alterations in the motor cortical and striatal glutamatergic system and D-serine levels in the bilateral 6-hydroxydopamine rat model for Parkinson's disease.
Alterations in the motor cortical and striatal glutamatergic system and D-serine levels in the bilateral 6-hydroxydopamine rat model for Parkinson's disease.
Neurochemistry international (2015-07-15)
Anissa El Arfani, Giulia Albertini, Eduard Bentea, Thomas Demuyser, Ann Van Eeckhaut, Ilse Smolders, Ann Massie
ZUSAMMENFASSUNG
Parkinson's disease (PD) is hallmarked by progressive degeneration of the substantia nigra pars compacta (SNc) neurons and is associated with aberrant glutamatergic activity. However, studies on the glutamatergic system in the motor cortex and striatum, two motor loop-related areas, are lacking in the clinically relevant bilateral SNc 6-hydroxydopamine (6-OHDA) rat model, and therefore led to the rationale behind the present investigations. Using Western blotting, the expression levels of the glial glutamate transporters, GLT-1 and GLAST, as well as xCT, the specific subunit of system xc(-), and the vesicular glutamate transporters, VGLUT1 and 2 were investigated at two different time points (1 week and 2 weeks) post-lesion. In addition, the total content of glutamate was measured. Moreover, the total D-serine levels were, to the best of our knowledge, studied for the first time in these two PD-related areas in the bilateral 6-OHDA rat model. In the motor cortex, no significant changes were observed in the different glutamate transporter expression levels in the bilaterally-lesioned rats. In the striatum, GLAST expression was significantly decreased at both time points whereas VGLUT1 and 2 expressions were significantly decreased 2 weeks after bilateral 6-OHDA lesion. Interestingly, bilateral 6-OHDA SNc lesion resulted in an enhancement of the total d-serine content in both motor cortex and striatum at 1 week post-lesion suggesting its possible involvement in the pathophysiology of PD. In conclusion, this study demonstrates disturbed glutamate and D-serine regulation in the bilateral SNc-lesioned brain which could contribute to the behavioral impairments in PD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
D-(+)-Glukose -Lösung, 45% in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumphosphat Dihydrat, BioUltra, Molecular Biology, ≥99.0% (T)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Phthaldialdehyd, ≥97% (HPLC), powder or crystals
Sigma-Aldrich
D-(+)-Glukose -Lösung, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Natriumbisulfat
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phthaldialdehyd, suitable for fluorescence, ≥99.0% (HPLC)
Sigma-Aldrich
α-D-Glucose, anhydrous, 96%
SAFC
BIS-TRIS
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Glucose -Lösung, BioUltra, Molecular Biology, ~20% in H2O
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Phthaldialdehyd, suitable for HPLC fluorimetric detection of amino acids, ≥99% (HPLC), powder or crystals