Merck
  • Lactobacillus pentosus var. plantarum C29 increases the protective effect of soybean against scopolamine-induced memory impairment in mice.
Lactobacillus pentosus var. plantarum C29 increases the protective effect of soybean against scopolamine-induced memory impairment in mice.
International journal of food sciences and nutrition (2015-07-15)
Dae-Hyoung Yoo, Dong-Hyun Kim
ZUSAMMENFASSUNG
Biological activities of soybean saponins are dependent on their metabolism by gut microbiota, which generate absorbable bioactive metabolites. Therefore, to enhance the pharmacological effect of soybean, we fermented defatted soybean powder (SP) with Lactobacillus pentosus var. plantarum C29 and measured its protective effect against scopolamine-induced memory impairment in mice using the passive avoidance, Y-maze and Morris water maze tasks. Fermentation increased soyasapogenol B, genistein and daidzein content of soybean and enhanced the protective effect of soybean against scopolamine-induced memory impairment. Additionally, compared with the exthanol extract of soybean, fermented SP (FSP) increased the expression of brain-derived neurotrophic factor (BDNF) in the hippocampi of scopolamine-treated mice. Furthermore, FSP inhibited acetylcholinesterase (AChE) activity in vitro and ex vivo. These findings suggest that C29 fermentation might increase the ameliorating effect of soybean against memory impairments by inhibiting AChE activity and increasing BDNF expression.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Genistein, synthetic, ≥98% (HPLC), powder
Sigma-Aldrich
(−)-Scopolamin -hydrobromid Trihydrat, >95% (HPLC), powder
Sigma-Aldrich
Daidzein, ≥98%, synthetic
Sigma-Aldrich
Scopolamin -hydrobromid, meets USP testing specifications
Sigma-Aldrich
Soyasaponin Bb
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Daidzin, ≥95.0% (HPLC)
Sigma-Aldrich
Genistin, from Glycine max (soybean), ≥95% (HPLC)
Sigma-Aldrich
Genistein, from Glycine max (soybean), ~98% (HPLC)