Merck
  • Simultaneous determination of scopolamine, hyoscyamine and anisodamine in in vitro growth media of selected Solanaceae hairy roots by CE method.
Simultaneous determination of scopolamine, hyoscyamine and anisodamine in in vitro growth media of selected Solanaceae hairy roots by CE method.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2015-08-09)
Szymon Dziomba, Teresa Łepek, Zbigniew Jaremicz, Maria Łuczkiewicz, Adam Prahl, Piotr Kowalski
ZUSAMMENFASSUNG
An electrophoretic method for fast separation of three tropane alkaloids (hyoscyamine, anisodamine and scopolamine) was presented. The substances were complete resolved in less than one minute due to utilization of relatively short capillary (20.2cm effective length) and high voltage (25kV). Detector probing frequency was found among the parameters that significantly affected the detection sensitivity. The performed experiments showed insufficient available probing frequency of used commercial spectrophotometric detector according to capillary electrophoresis (CE) separation potential. Under the optimized conditions the background electrolyte (BGE) was composed of 20mM Tris, 6mM HCl and 20mM NaCl (pH 8.50). All analyses were carried out in fused silica capillaries of 50μm (inner diameter) and 31.2cm (total capillary length). Samples were injected hydrodynamically (5s; 3.45kPa) without any sample preparation step and separation was performed at 25kV. The elaborated method was applied in plant cultures growth media analysis after incubation with hairy roots of selected Solanaceae species. The performed experiments proved the usefulness of CE in quality control of biotechnological processes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Trizma® Base, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
(−)-Scopolamin -hydrobromid Trihydrat, >95% (HPLC), powder
Sigma-Aldrich
Scopolamin -hydrobromid, meets USP testing specifications
Sigma-Aldrich
Trizma® Base, 99.7% (T), puriss. p.a.
SAFC
Tromethamin