Merck
  • Development of fluorinated methoxylated chalcones as selective monoamine oxidase-B inhibitors: Synthesis, biochemistry and molecular docking studies.
Development of fluorinated methoxylated chalcones as selective monoamine oxidase-B inhibitors: Synthesis, biochemistry and molecular docking studies.
Bioorganic chemistry (2015-07-21)
Bijo Mathew, Githa Elizabeth Mathew, Gülberk Uçar, Ipek Baysal, Jerad Suresh, Jobin Kunjumon Vilapurathu, Aneesh Prakasan, Jeethu Kuruppath Suresh, Anjana Thomas
ZUSAMMENFASSUNG
A series of methoxylated chalcones with fluoro and trifluoromethyl derivatives were synthesized and investigated for their ability to inhibit human monoamine oxidase A and B. The chemical structures of the compounds have been characterized by means of their (1)H NMR, (13)C NMR, Mass spectroscopic datas and elemental analysis. The results demonstrate that these compounds are reversible and selective MAO-B inhibitors with a competitive mode of inhibition. The most potent compound (2E)-1-(4-methoxyphenyl)-3-[4-(trifluoromethyl)phenyl] prop-2-en-1-one showed the best activity and higher selectivity towards hMAO-B with Ki and SI values of 0.22±0.01μM and 0.05 comparable to that standard drug, Selegiline Ki and SI values were found as 0.33±0.03μM and 0.04, respectively. Molecular docking studies were carried out to further explain the in vitro results of the new compounds, and to identify the hypothetical binding mode for the compounds inside the inhibitor binding cavity of hMAO-B.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Salzsäure -Lösung, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
[Bis-(trifluoracetoxy)-iod]-benzol, 97%
Sigma-Aldrich
Salzsäure -Lösung, 32 wt. % in H2O, FCC
Sigma-Aldrich
Resorufin, 95%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®