Merck
  • In Vitro Validation of Survivin as Target Tumor-associated Antigen for Immunotherapy in Uterine Cancer.
In Vitro Validation of Survivin as Target Tumor-associated Antigen for Immunotherapy in Uterine Cancer.
Journal of immunotherapy (Hagerstown, Md. : 1997) (2015-06-08)
Anke Vanderstraeten, Tina Everaert, Rieta Van Bree, Godelieve Verbist, Cathérine Luyten, Frederic Amant, Sandra Tuyaerts
ZUSAMMENFASSUNG
Survivin is an antiapoptotic protein, not expressed in terminally differentiated adult tissues, yet overexpressed in several tumors. Therefore, it is an interesting target for immunotherapeutic strategies. In addition to specific overexpression in tumors, tumor survival is mediated by survivin and hence, tumor survival can be tackled by targeting survivin. Survivin expression in uterine cancer was validated by quantitative real-time polymerase chain reaction and immunohistochemistry. In addition, we evaluated survivin immunogenicity by analyzing spontaneous B-cell and T-cell responses in patients. Survivin as a protein was expressed in only a minority of normal tissues, whereas it was being expressed in all of the currently analyzed uterine cancers, both endometrial carcinoma (n = 52) and uterine sarcoma (n = 52). Survivin RNA transcripts were overexpressed in more aggressive tumors and survivin protein was overexpressed in recurrent endometrial tumors compared with primary tumors. Spontaneous T-cell responses were seen in 10/39 endometrial cancer patients and 3/16 uterine sarcoma patients. In normal controls, T-cell responses were found only in 1 donor (n = 21). Although increased antibody titers were found in more aggressive and far-advanced tumors, no differences in B-cell responses were seen. Overall, when compared with normal controls, a B-cell response was only measured in 1/41 uterine sarcoma patients. In conclusion, we currently validated the presence of survivin in uterine cancer. In addition, spontaneous T-cell responses were found in 23.6% of the total patient population. These data indicate that a survivin-specific immune response may be induced spontaneously in patients, further fortifying the eligibility of survivin as an immunotherapeutic target.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®