Merck
  • Effects of Dietary Brazilian Palm Oil (Mauritia flexuosa L.) on Cholesterol Profile and Vitamin A and E Status of Rats.
Effects of Dietary Brazilian Palm Oil (Mauritia flexuosa L.) on Cholesterol Profile and Vitamin A and E Status of Rats.
Molecules (Basel, Switzerland) (2015-05-23)
Jailane de Souza Aquino, Juliana Késsia Barbosa Soares, Marciane Magnani, Thayza Christina Montenegro Stamford, Robson de Jesus Mascarenhas, Renata Leite Tavares, Tânia Lúcia Montenegro Stamford
ZUSAMMENFASSUNG
In vitro studies have been carried out to establish the nutritional differences between crude and refined vegetable oils; however, the impact of the consumption of these foods on metabolism, in particular the effect of buriti oil, needs to be further evaluated. The aim of this study was to evaluate the biochemical and murine parameters and the vitamin A and E status in young rats fed with diets supplemented with crude or refined buriti oil. The animals (n = 30) were randomized into three groups receiving diet added of soybean oil (control), crude buriti oil (CBO) and refined buriti oil (RBO) for 28 days. Rats fed with diet added of refined buriti oil (RBO) showed reduced total cholesterol (up to 60.27%), LDL (64.75%), triglycerides (55.47%) and enzyme aspartate transaminase (21.57%) compared to those fed with diet added of crude oil. Serum and hepatic retinol and tocopherol were higher by two to three times in CBO and RBO groups compared to the control group, but no differences were observed for murine parameters. The results indicate that buriti oil is an important source of the antioxidant vitamins A and E, and refined buriti oil is suggested as alternative to improve the lipid profile of healthy rats.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.