Merck
  • Aquaporin 1 and 5 expression decreases during human intervertebral disc degeneration: Novel HIF-1-mediated regulation of aquaporins in NP cells.
Aquaporin 1 and 5 expression decreases during human intervertebral disc degeneration: Novel HIF-1-mediated regulation of aquaporins in NP cells.
Oncotarget (2015-04-07)
Zariel I Johnson, Shilpa S Gogate, Rebecca Day, Abbie Binch, Dessislava Z Markova, Neil Chiverton, Ashley Cole, Matt Conner, Irving M Shapiro, Christine L Le Maitre, Makarand V Risbud
ZUSAMMENFASSUNG
Objectives of this study were to investigate whether AQP1 and AQP5 expression is altered during intervertebral disc degeneration and if hypoxia and HIF-1 regulate their expression in NP cells. AQP expression was measured in human tissues from different degenerative grades; regulation by hypoxia and HIF-1 was studied using promoter analysis and gain- and loss-of-function experiments. We show that both AQPs are expressed in the disc and that mRNA and protein levels decline with human disease severity. Bioinformatic analyses of AQP promoters showed multiple evolutionarily conserved HREs. Surprisingly, hypoxia failed to induce promoter activity or expression of either AQP. While genomic chromatin immunoprecipitation showed limited binding of HIF-1α to conserved HREs, their mutation did not suppress promoter activities. Stable HIF-1α suppression significantly decreased mRNA and protein levels of both AQPs, but HIF-1α failed to induce AQP levels following accumulation. Together, our results demonstrate that AQP1 and AQP5 expression is sensitive to human disc degeneration and that HIF-1α uniquely maintains basal expression of both AQPs in NP cells, independent of oxemic tension and HIF-1 binding to promoter HREs. Diminished HIF-1 activity during degeneration may suppress AQP levels in NP cells, compromising their ability to respond to extracellular osmolarity changes.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
3,3′-Diaminobenzidin -tetrahydrochlorid, tablet, 10 mg substrate per tablet
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
DMOG, ≥98% (HPLC)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis