Merck
  • Hygroscopic and phase transition properties of alkyl aminium sulfates at low relative humidities.
Hygroscopic and phase transition properties of alkyl aminium sulfates at low relative humidities.
Physical chemistry chemical physics : PCCP (2015-07-15)
Yangxi Chu, Meike Sauerwein, Chak K Chan
ZUSAMMENFASSUNG
Alkyl aminium sulfates (AASs) can affect the physicochemical properties of atmospheric aerosols such as hygroscopicity. Previous laboratory experiments have shown that the water content in AAS bulk solutions is higher than in aqueous ammonium sulfate solution in the range of 60-95% relative humidity (RH). Furthermore, amine was found to evaporate from the solution during the preparation of AASs from the parent amine and sulfuric acid solutions. Here we report the hygroscopicities of deposited particles of four AASs at different aminium-to-sulfate molar ratios (A/Ss) in the range of <3-90% RH using air-flow cells coupled with in situ micro-Raman spectroscopy. Normalized integrated areas of O-H stretching peaks in the Raman spectra were converted to water-to-solute molar ratios (WSRs) at various RH values. Evaporation of amine was also observed in most cases and the exact A/Ss of sample particles or solutions were determined by ion chromatography. Mono-methylaminium sulfate (MMAS) and mono-ethylaminium sulfate (MEAS) particles were stable at A/S = 2.0, but di-methylaminium sulfate (DMAS) and tri-methylaminium sulfate (TMAS) suffered from DMA and TMA evaporation and eventually equilibrated to the A/S of 1.5 and 1.0, respectively. At these stable compositions MMAS and MEAS exhibited phase transitions in the super-saturation region, while DMAS and TMAS showed a continuous and reversible water uptake. Besides, an approach to estimate the hygroscopicities of DMAS and TMAS particles at an initial A/S larger than that of the stable compositions was presented. In the range of 60-95% RH, the WSRs of all the studied AAS particles were consistent with a previous study based on experimental values and the extended Zdanovskii-Stokes-Robinson equation. In general, all the studied AASs were more hygroscopic than their corresponding ammonium counterparts within the studied RH range and evaporation of amine needs to be corrected in studying unstable AAS particles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
Sigma-Aldrich
Trimesinsäure, 95%
Sigma-Aldrich
Ethanolamin, ≥99%
Sigma-Aldrich
Methylamin -Lösung, 33 wt. % in absolute ethanol ((denatured with 1% toluene))
Sigma-Aldrich
Methylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Ethanolamin, ≥98%
Sigma-Aldrich
Dimethylamin -Lösung, 40 wt. % in H2O
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in toluene
Sigma-Aldrich
Ethanolamin, purified by redistillation, ≥99.5%
Sigma-Aldrich
Dimethylamin -Lösung, 2.0 M in THF
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Dimethylamin, anhydrous, ≥99%
Sigma-Aldrich
Trimethylamin -Lösung, 43.0-49.0% in H2O (T)
Sigma-Aldrich
Trimethylaluminium, 97%
Sigma-Aldrich
Ethanolamin, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Trimethylamin -Lösung, 31-35 wt. % in ethanol, 4.2 M, contains toluene as stabilizer
Sigma-Aldrich
Methylamin -Lösung, 2.0 M in methanol
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Trimethylamin -Lösung, 25 wt. % in H2O
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Trimethylaluminium, packaged for use in deposition systems
Sigma-Aldrich
Trimethylaluminium -Lösung, 2.0 M in hexanes