Merck
  • Cyclin-dependent kinase 5 regulates the proliferation, motility and invasiveness of lung cancer cells through its effects on cytoskeletal remodeling.
Cyclin-dependent kinase 5 regulates the proliferation, motility and invasiveness of lung cancer cells through its effects on cytoskeletal remodeling.
Molecular medicine reports (2015-05-29)
Jun-Li Liu, Run-Xia Gu, Xiao-Shu Zhou, Fang-Zheng Zhou, Gang Wu
ZUSAMMENFASSUNG
Determining the molecular phenotype is a key to understanding and predicting the metastatic potential and the prognosis for patients with lung cancer. Our previous study demonstrated that increased expression of cyclin‑dependent kinase 5 (CDK5) in patients with non‑small cell lung cancer (NSCLC) is associated with a poorer prognosis. The present study aimed to further investigate the underlying mechanism of CDK5 in vitro and in vivo using the A549 human NSCLC cell line. A 3‑(4,5‑dimethylthiazol‑2‑yl)‑2,5‑diphenyltetrazolium bromide assay was used to quantify the proliferation of the A549 cells; migration assay and invasiveness assays were performed using Transwell chambers and wound healing assays were used to assess cell motility, which was assessed by measuring the movement of cells. Inhibition of CDK5 by roscovitine and small interfering (si)RNA was used to investigate the mechanism of CDK5 in the process of A549 lung cancer cell proliferation, migration and invasion. The results demonstrated that functional inhibition of CDK5 using roscovitine and siRNA markedly suppressed the proliferation of A549 cells and resulted in a reduced tumor mass in vivo. In addition, the hinhibition of CDK5 reduced the migration and invasiveness of the A549 cells in vitro and in vivo. Notably, CDK5 inhibition also impaired tumor cell cytoskeletal remodeling and led to loss of cell polarity, which may partially explain the reduction of A549 cell mobility and invasiveness. The results of the present study revealed that CDK5 may be important in the regulation of migration and invasiveness in NSCLC through its effects on cytoskeletal remodeling.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, BioReagent, suitable for fluorescence, mixture of 2 components, ≥90% (HPLC)
Sigma-Aldrich
Fluoreszein-5(6)-isothiocyanat, ≥90% (HPLC)
Sigma-Aldrich
Fluorescein-Isothiocyanat Isomer I, ≥97.5% (HPLC)