Merck
  • Oxidovanadium(IV) sulfate-induced glucose uptake in HepG2 cells through IR/Akt pathway and hydroxyl radicals.
Oxidovanadium(IV) sulfate-induced glucose uptake in HepG2 cells through IR/Akt pathway and hydroxyl radicals.
Journal of inorganic biochemistry (2015-05-30)
Qian Zhao, Deliang Chen, Pingsheng Liu, Taotao Wei, Fang Zhang, Wenjun Ding
ZUSAMMENFASSUNG
The insulin-mimetic and anti-diabetic properties of vanadium and related compounds have been well documented both in vitro and in vivo. However, the molecular basis of the link between vanadium and the insulin signaling pathway in diabetes mellitus is not fully described. We investigated the effects of reactive oxygen species (ROS) induced by oxidovanadium(IV) sulfate (VOSO4) on glucose uptake and the insulin signaling pathway in human hepatoma cell line HepG2. Exposure of cells to VOSO4 (5-50 μM) resulted in an increase in glucose uptake, insulin receptor (IR) and protein kinase B (Akt) phosphorylation and intracellular ROS generation. Using Western blot, we found that catalase and sodium formate, but not superoxide dismutase, prevented the increase of hydroxyl radical (·OH) generation and significantly decreased VOSO4-induced IR and Akt phosphorylation. These results suggest that VOSO4-induced ·OH radical, which is a signaling species, promotes glucose uptake via the IR/Akt signaling pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
2′,7′-Dichlorfluorescein-Diacetat, ≥97%
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Rotenon, ≥95%
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Diphenyleniodoniumchlorid, ≥98%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Natriumformiat, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumformiat, 99.998% trace metals basis
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri