Merck
  • Anti-oxidative responses of zebrafish (Danio rerio) gill, liver and brain tissues upon acute cold shock.
Anti-oxidative responses of zebrafish (Danio rerio) gill, liver and brain tissues upon acute cold shock.
Comparative biochemistry and physiology. Part A, Molecular & integrative physiology (2015-05-31)
Su Mei Wu, Jia-Hao Liu, Li-Hsin Shu, Ching Hsein Chen
ZUSAMMENFASSUNG
The present study seeks to detect oxidative damage and to compare anti-oxidative responses among liver, gills and brain of adult zebrafish that were cooled from 28 °C (control) to 12 °C (treatment) for 0-24 h. The lipid peroxidation of liver, gill and brain tissues significantly increased at 1h after transfer, but reactive oxygen species in the treatment group increased significantly after 24 h as compared to the control. The fish were found to develop a cascading anti-oxidative mechanism beginning with an increase in Cu/Zn-SOD levels, followed by increased CAT and GPx mRNA expressions in the three tissue types. Both smtB and mt2 mRNAs increased in the hepatic and brain tissues following 1h of cold stress, but only smtB exhibited a significant increase in the gills at 1 h and 6 h after transfer to 12 °C. Furthermore, cellular apoptosis in the brain was not evident after cold shock, but liver and gills showed cellular apoptosis at 1-3 h, with another peak in the liver at 6 h after cold shock. The results suggest that the cold shock induced oxidative stress, and the enzymatic (SOD, GPx and CAT) and non-enzymatic (mt-2 and smt-B) mRNA expressions all play a role in the resulting anti-oxidation within 1-6 h of cold shock. A functional comparison showed that the brain had the most powerful antioxidant defense system of the three tissue types since it had the highest smtB mRNA expression and a lower level of cell apoptosis than the liver and gills after exposure to cold stress.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Hämatoxylin
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Hämatoxylin, certified by the BSC
Sigma-Aldrich
o-Xylen, anhydrous, 97%
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis