Merck
Cdc42 and sec10 Are Required for Normal Retinal Development in Zebrafish.
Investigative ophthalmology & visual science (2015-05-30)
Soo Young Choi, Jeong-In Baek, Xiaofeng Zuo, Seok-Hyung Kim, Joshua L Dunaief, Joshua H Lipschutz
ZUSAMMENFASSUNG
To characterize the function and mechanisms of cdc42 and sec10 in eye development in zebrafish. Knockdown of zebrafish cdc42 and sec10 was carried out using antisense morpholino injection. The phenotype of morphants was characterized by histology, immunohistology, and transmission electron microscopy (TEM). To investigate a synergistic genetic interaction between cdc42 and sec10, we titrated suboptimal doses of cdc42 and sec10 morpholinos, and coinjected both morpholinos. To study trafficking, a melanosome transport assay was performed using epinephrine. Cdc42 and sec10 knockdown in zebrafish resulted in both abnormal eye development and increased retinal cell death. Cdc42 morphants had a relatively normal retinal structure, aside from the absence of most connecting cilia and outer segments, whereas in sec10 morphants, much of the outer nuclear layer, which is composed of the photoreceptor nuclei, was missing and RPE cell thickness was markedly irregular. Knockdown of cdc42 and sec10 also resulted in an intracellular transport defect affecting retrograde melanosome transport. Furthermore, there was a synergistic genetic interaction between zebrafish cdc42 and sec10, suggesting that cdc42 and sec10 act in the same pathway in retinal development. We propose a model whereby sec10 and cdc42 play a central role in development of the outer segment of the retinal photoreceptor cell by trafficking proteins necessary for ciliogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Proteasehemmer-Cocktailpulver, for general use, lyophilized powder
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Monoklonales Anti-Tubulin, acetyliert in Maus hergestellte Antikörper, clone 6-11B-1, ascites fluid
Sigma-Aldrich
Phenol red solution, 0.5%, liquid, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
Phenolrot, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Phenolrot, ACS reagent
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
Natriumdodecylsulfat, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
Natriumdodecylsulfat, ≥90% ((Assay))
Sigma-Aldrich
Natriumdodecylsulfat, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri