Merck
  • Chemical Characterization of N-Linked Oligosaccharide As the Antigen Epitope Recognized by an Anti-Sperm Auto-Monoclonal Antibody, Ts4.
Chemical Characterization of N-Linked Oligosaccharide As the Antigen Epitope Recognized by an Anti-Sperm Auto-Monoclonal Antibody, Ts4.
PloS one (2015-07-30)
Hiroshi Yoshitake, Noritaka Hashii, Nana Kawasaki, Shuichiro Endo, Kenji Takamori, Akiko Hasegawa, Hiroshi Fujiwara, Yoshihiko Araki
ZUSAMMENFASSUNG
Ts4, an anti-sperm auto-monoclonal antibody, possesses immunoreactivity to the acrosomal region of mouse epididymal spermatozoa. In addition, the mAb shows specific immunoreactivity to reproduction-related regions such as testicular germ cells and early embryo. Our qualitative study previously showed that the antigen epitope for Ts4 contained a N-linked common oligosaccharide (OS) chain on testicular glycoproteins as determined by Western blotting for testicular glycoproteins after treatment with several glycohydrolases. Since the distribution of the Ts4-epitope is unique, the OS chain in Ts4-epitope may have role(s) in the reproductive process. The aim of this study was to clarify the molecular structure of the Ts4-epitope, particularly its OS moiety. Using Ts4 immunoprecipitation combined with liquid chromatography and multiple-stage mass spectrometry, the candidate carbohydrate structure in the Ts4-epitope is proposed to be N-linked fucosylated agalacto-biantennary with bisecting N-acetylglucosamine (GlcNAc) or with N-acetylgalactosamine-GlcNAc motif. Further binding analyses using various lectins against the mouse testicular Ts4-immunoprecipitants revealed that Phaseolus vulgaris erythroagglutinin and Pisum sativum agglutinin showed positive staining of the bands corresponding to Ts4 reactive proteins. Moreover, the immunoreactivity of Ts4 against the testicular extract was completely abrogated after digestion with β-N-acetylglucosaminidase. These results show that the Ts4-epitope contains agalacto-biantennary N-glycan with bisecting GlcNAc carrying fucose residues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Kohlenstoff, mesoporös, less than 100 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, less than 500 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
Kohlenstoff, mesoporös
Sigma-Aldrich
Kohlenstoff, mesoporös, nanopowder, graphitized, less than 250 ppm Al, Ti, Fe, Ni, Cu, and Zn combined
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, purum, ≥99.0% (TLC)
Sigma-Aldrich
Methan-12C, 99.99 atom % 12C
Carbon - Vitreous, rod, 100mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 2.5mm, bulk density 0.05g/cm3, porosity 96.5%
Sigma-Aldrich
Essigsäure-12C2, 99.9 atom % 12C
Carbon - Vitreous, foil, 25x25mm, thickness 0.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, rod, 5mm, diameter 3.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, thickness 3.2mm, bulk density 0.05g/cm3, porosity 96.5%
Carbon - Vitreous, tube, 100mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, rod, 50mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, foam, 150x150mm, 0.05g.cmué, porosity 96.5%, 24 pores/cm
Carbon - Vitreous, foil, 50x50mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 1.0mm, glassy carbon
Carbon - Vitreous, rod, 200mm, diameter 5.0mm, glassy carbon
Carbon - Vitreous, foil, 10x10mm, thickness 4.0mm, glassy carbon
Carbon - Vitreous, tube, 50mm, outside diameter 10mm, inside diameter 3mm, wall thickness 3.5mm, glassy carbon
Carbon - Vitreous, rod, 100mm, diameter 1.0mm, glassy carbon