Merck
  • Biochemical features and antiviral activity of a monomeric catalytic antibody light-chain 23D4 against influenza A virus.
Biochemical features and antiviral activity of a monomeric catalytic antibody light-chain 23D4 against influenza A virus.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2015-02-26)
Emi Hifumi, Mitsue Arakawa, Shingo Matsumoto, Tatsuhiro Yamamoto, Yoshiki Katayama, Taizo Uda
ZUSAMMENFASSUNG
Catalytic antibodies have exhibited interesting functions against some infectious viruses such as HIV, rabies virus, and influenza virus in vitro as well as in vivo. In some cases, a catalytic antibody light chain takes on several structures from the standpoint of molecular size (monomer, dimer, etc.) and/or isoelectronic point. In this study, we prepared a monomeric 23D4 light chain by mutating the C-terminal Cys to Ala of the wild-type. The mutated 23D4 molecule took a simple monomeric form, which could hydrolyze synthetic 4-methyl-coumaryl-7-amide substrates and a plasmid DNA. Because the monomeric 23D4 light chain suppressed the infection of influenza virus A/Hiroshima/37/2001 in an in vitro assay, the corresponding experiments were conducted in vivo, after the virus strain (which was taken from a human patient) was successfully adapted into BALB/cN Sea mice. In the experiments, a mixture of the monomeric 23D4 and the virus was nasally administered 1) with preincubation and 2) without preincubation. As a result, the monomeric 23D4 clearly exhibited the ability to suppress the influenza virus infection in both cases, indicating a potential drug for preventing infection of the influenza A virus.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
6-Maleimidohexansäure, 90% (GC)
Sigma-Aldrich
Ampicillin, meets USP testing specifications
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis