Merck
Kinetics of phytosterol metabolism in neonates receiving parenteral nutrition.
Pediatric research (2015-04-22)
T Hang Nghiem-Rao, Ilker Tunc, Alisha M Mavis, Yumei Cao, Elizabeth M Polzin, Mary F Firary, Xujing Wang, Pippa M Simpson, Shailendra B Patel
ZUSAMMENFASSUNG
Phytosterols in soybean oil (SO) lipids likely contribute to parenteral nutrition-associated liver disease (PNALD) in infants. No characterization of phytosterol metabolism has been done in infants receiving SO lipids. In a prospective cohort study, 45 neonates (36 SO lipid vs. 9 control) underwent serial blood sample measurements of sitosterol, campesterol, and stigmasterol. Mathematical modeling was used to determine pharmacokinetic parameters of phytosterol metabolism and phytosterol exposure. Compared to controls, SO lipid-exposed infants had significantly higher levels of sitosterol and campesterol (P < 0.01). During SO lipid infusion, sitosterol and campesterol reached half of steady-state plasma levels within 1.5 and 0.8 d, respectively. Steady-state level was highest for sitosterol (1.68 mg/dl), followed by campesterol (0.98 mg/dl), and lowest for stigmasterol (0.01 mg/dl). Infants born < 28 wk gestational age had higher sitosterol steady-state levels (P = 0.03) and higher area under the curve for sitosterol (P = 0.03) during the first 5 d of SO lipid (AUC5) than infants born ≥ 28 wk gestational age. Phytosterols in SO lipid accumulate rapidly in neonates. Very preterm infants receiving SO lipid have higher sitosterol exposure, and may have poorly developed mechanisms of eliminating phytosterols that may contribute to their vulnerability to PNALD.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
β-Sitosterol, synthetic, ≥95%
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Stigmasterol, ~95%
Sigma-Aldrich
β-Sitosterol, ≥70%
Sigma-Aldrich
Campesterol, ~65%
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
β-Sitosterol, from soybean, ≥96%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications