Merck
  • Periaortic Fat Tissue: A Predictor of Cardiac Valvular Calcification, Malnutrition, Inflammation, and Atherosclerosis Components in Hemodialysis Patients.
Periaortic Fat Tissue: A Predictor of Cardiac Valvular Calcification, Malnutrition, Inflammation, and Atherosclerosis Components in Hemodialysis Patients.
Artificial organs (2015-05-06)
Gultekin Genctoy, Olcay Eldem, Tarkan Ergun, Serap Arikan
ZUSAMMENFASSUNG
Cardiac valvular calcification (CVC) in end-stage renal disease is shown to be a component of malnutrition, inflammation, atherosclerosis, calcification (MIAC) syndrome. Thoracic periaortic fat tissue (T-PAFT) is shown to be increased in patients with end-stage renal disease (ESRD), and has positive correlation with MIAC. Negative correlation between CVC and vitamin D is shown in hemodialysis (HD) patients. In this study, we investigated a relationship between body composition, T-PAFT, metabolic and inflammatory parameters, and CVC in HD patients. Seventy-six HD patients (49M) were included. CVC is defined as bright echoes of >1 mm on one or more cusps on echocardiography. Results were expressed as the number of calcified valves (0,1,2). Calcium, phosphorus, parathyroid hormone (PTH), C-reactive protein (CRP), albumin and 25-hydroxy vitamin D levels were studied from predialysis blood samples. T-PAFT was calculated using a method with manual definition of borders on images from multislice computed tomography. Basal metabolic rate, muscle mass, total and truncal fat mass were measured by bioimpedance analysis. There were 65.8% of patients who had CVC. Patients with CVC were older (63.5 ± 14.6 ± 17, P = 0.02). T-PAFT (1599 ± 596, 739.7 ± 179 mm(2) , P = 0.001) and CRP (15.8 ± 11; 11.1 ± 13.2 mg/dL; P = 0.04) were higher in the group with CVC. T-PAFT had positive correlations with CRP, MIAC, body mass index (BMI) and number of calcified valves, negative correlation with left ventricular ejection fraction, and no correlation with albumin, calcium, phosphorus, and PTH. The logistic regression analysis revealed that T-PAFT was a significant predictor of CVC. In this study, T-PAFT showed a positive correlation with inflammation, CVC, and MIAC score in HD patients. T-PAFT was a significant predictor of CVC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications