Merck
  • Cysticidal activity of extracts and isolated compounds from Teloxys graveolens: In vitro and in vivo studies.
Cysticidal activity of extracts and isolated compounds from Teloxys graveolens: In vitro and in vivo studies.
Experimental parasitology (2015-06-15)
Francisca Palomares-Alonso, Irma Susana Rojas-Tomé, Victorino Juárez Rocha, Guadalupe Palencia Hernández, Angélica González-Maciel, Andrea Ramos-Morales, Rosalba Santiago-Reyes, Iliana Elvira González-Hernández, Helgi Jung-Cook
ZUSAMMENFASSUNG
In the search of new alternatives for neurocysticercosis treatment, the cysticidal activity of organic extracts of Teloxys graveolens was evaluated. The in vitro activity of hexane, ethyl acetate and methanol extracts against Taenia crassiceps cysts was tested and the selectivity index relative to human fibroblasts was determined. Subsequently, the in vivo efficacy of the methanolic extract at doses of 200 and 500 mg/kg in the murine cysticercosis model was evaluated. The ultrastructural effects in vitro and in vivo of the methanolic extract were also investigated using scanning electron microscopy. Additionally, a bioassay-guided fractionation for the isolation of the cysticidal components was performed. Our in vitro findings revealed that all extracts exhibited good cysticidal activity with EC50 values from 44.8 to 67.1 µg/mL. Although the ethyl acetate and methanolic extracts displayed low cytotoxicity, the methanolic extract was the most selective. The methanolic extract also showed in vivo efficacy which was similar to that obtained with ABZ. Significant alterations were found on the germinal layer of the cysts, with a high accumulation of granules of glycogen and vacuoles. The bioguided fractionation of methanolic extract led to the isolation of three flavonoids: chrysin, pinocembrin and pinostrobin; among them, pinocembrin was the compound that displayed cysticidal activity. This is the first study which reveals that T. graveolens could be a potential source for cysticidal and non-toxic compounds.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Schwefelsäure, 99.999%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Osmiumtetroxid, Sealed ampule.
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 4% in H2O
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Osmiumtetroxid -Lösung, suitable for electron microscopy, 2% in H2O
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Os EnCat® 40, extent of labeling: 0.3 mmol/g Os loading
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)