Merck
  • Radiosynthesis of 4-[(18)F]fluoro-L-tryptophan by isotopic exchange on carbonyl-activated precursors.
Radiosynthesis of 4-[(18)F]fluoro-L-tryptophan by isotopic exchange on carbonyl-activated precursors.
Bioorganic & medicinal chemistry (2015-07-22)
Philipp S Weiss, Johannes Ermert, Johnny Castillo Meleán, Dominique Schäfer, Heinz H Coenen
ZUSAMMENFASSUNG
Several (18)F-labeled aromatic amino acids have been developed primarily for tumor imaging with positron-emission-tomography (PET). Also, (18)F-labeled tryptophan derivatives were synthesized by electrophilic (18)F-fluorination or by introducing a [(18)F]fluoroalkyl group. Here, a 3-step method for a nucleophilic radiosynthesis of 4-[(18)F]fluoro-L-tryptophan was developed. A carbonyl activated precursor containing a chiral amino acid building block was radiofluorinated by isotopic exchange, followed by removal of the activating formyl group by reductive decarbonylation and subsequent cleavage of the building block under acidic conditions. The title compound was obtained within 100 min with a radiochemical yield of about 13%, a molar activity of >70 MBq/mmol and an enantiomeric excess of >99%.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis