Merck
  • Synthesis and characterization of composite polymer, polyethylene glycol grafted flower-like cupric nano oxide for solid phase microextraction of ultra-trace levels of benzene, toluene, ethyl benzene and o-xylene in human hair and water samples.
Synthesis and characterization of composite polymer, polyethylene glycol grafted flower-like cupric nano oxide for solid phase microextraction of ultra-trace levels of benzene, toluene, ethyl benzene and o-xylene in human hair and water samples.
Journal of chromatography. A (2015-09-29)
Ali Sarafraz-Yazdi, Amene Zendegi-Shiraz, Zarrin Es'haghi, Mohammad Hassanzadeh-Khayyat
ZUSAMMENFASSUNG
In this research, poly (ethylene glycol)-poly (ethylene glycol) grafted flower-like cupric oxidenano particles (PEG-PEG-g-CuO NPs) as a novel fiber coating of solid-phase microextraction (SPME) were synthesized by using sol-gel technology. This fiber was successfully applied to extract and determine the ultra-trace levels of benzene, toluene, ethyl benzene and o-xylene in human hair using head space-solid-phase microextraction (HS-SPME) coupled to gas chromatography-flame ionization detector. Characterization and chemical composition of the nano particle was performed by Fourier transform infrared spectroscopy (FT-IR), energy dispersion spectroscopy (EDS) and back scatter analysis (BSA). These methods confirmed the successful fabrication of PEG-g-CuO NPs. The surface morphology of the fibers were inspected by scanning electron microscopy. The scanning electron microscopy (SEM) revealed many "crack-like" features and highly porous structure on the surface of fiber. The synthesized nanocomposites were used for preconcentration and extraction of benzene, toluene, ethyl benzene and o-xylene (BTEX). The effects of operating parameters such as: desorption temperature and time, extraction temperature, extraction time, stirring speed and salt effect were investigated and optimized. Under the optimal conditions, the method detection limits and the limits of quantification were between 0.00025-50.00000pgmL(-1) and 0.00200-200.00000pgmL(-1), respectively. Linearity was observed over a range 0.00200-200000.00000pgmL(-1). The relative standard deviations for one fiber (repeatability; n=5) were obtained from 3.30 up to 5.01% and between fibers or batch to batch (n=3; reproducibility) in the range of 3.63-6.21%. The developed method was successfully applied to simultaneous determination of BTEX in human hairs, tap water and distillate water.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Sigma-Aldrich
Trifluoressigsäure, ReagentPlus®, 99%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenglycol, ReagentPlus®, ≥99%
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Trifluoressigsäure, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenglycol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Trifluoressigsäure, ≥99%, for protein sequencing
Sigma-Aldrich
Ethylenglycol, spectrophotometric grade, ≥99%
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1 cm), needle size 23 ga, Autosampler, pk of 3, plain gray hub
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
o-Xylen, reagent grade, ≥98.0%
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (2cm), needle size 24 ga, Autosampler, pk of 3, notched gray hub
Sigma-Aldrich
Aceton, ACS reagent, ≥99.5%
Supelco
SPME-Faseranordnung, 50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)