Merck
  • The decomposition of protoporphyrin IX by ultrasound is dependent on the generation of hydroxyl radicals.
The decomposition of protoporphyrin IX by ultrasound is dependent on the generation of hydroxyl radicals.
Ultrasonics sonochemistry (2015-05-03)
Haobo Xu, Xin Sun, Jianting Yao, Jian Zhang, Yun Zhang, Haibo Chen, Juhua Dan, Zhen Tian, Ye Tian
ZUSAMMENFASSUNG
The ultrasound activation of certain drugs, such as porphyrins, could cause synergistic cytotoxic effects on cells. Both sonomechanical and sonochemical effects occur and the latter play a critical role because antioxidant agents could exert significant protective effects against the cytotoxicity. To investigate the reactive oxygen species involved in the sonochemical effects, aqueous protoporphyrin IX (PpIX) solutions were characterized under ultrasound sonication in this study. Inertial cavitation was indirectly evaluated using terephthalic acid dosimetry. The fluorescence intensity of the PpIX was measured using a fluorescence spectrophotometer. The effects of PpIX concentration, ultrasound parameters and free radical scavengers on the PpIX activation by ultrasound were investigated. Our results showed that the increase in PpIX decomposition was significantly correlated with cavitation activities (R=0.9874, p<0.05), and the decomposing effect increases with ultrasound intensity (0.6-1.5 W/cm(2)), initial PpIX concentration (1-5 μM), duty cycle (10-100%) and the sonication duration (2-10 min). The fluorescence and absorption spectra of PpIX showed a decrease in the peak intensity without spectral shifts or new peak build-up after sonication. The PpIX decomposition was significantly inhibited by hydroxyl radical scavengers, histidine, mannitol, acetone, methanol and ethanol, but the decomposition was not inhibited by sodium azide, catalase or superoxide dismutase. These results suggest that the decomposition of protoporphyrin IX by ultrasound is dependent on the generation of hydroxyl radicals, which sheds some light on the sonochemical effects of the interaction between ultrasound and porphyrins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Natriumazid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Calciumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
Calciumchlorid, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Terephthalsäure, 98%
Sigma-Aldrich
Natriumazid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
HEPES-Pufferlösung, 1 M in H2O
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Natriumazid, purum p.a., ≥99.0% (T)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Natriumazid, BioXtra
Sigma-Aldrich
Calciumchlorid, powder, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid Dihydrat
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Calciumchlorid, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis