Merck
1,8-Cineole Ameliorates Steatosis of Pten Liver Specific KO Mice via Akt Inactivation.
International journal of molecular sciences (2015-05-30)
Soichiro Murata, Koichi Ogawa, Takashi Matsuzaka, Mitsuru Chiba, Ken Nakayama, Kenichi Iwasaki, Tomohiro Kurokawa, Naoki Sano, Tomohito Tanoi, Nobuhiro Ohkohchi
ZUSAMMENFASSUNG
Hepatocyte-specific Phosphatase and tensin homolog (Pten)-knockout (KO) mice exhibit hepatic lesions analogous to non-alcoholic steatohepatitis (NASH). 1,8-cineole is a monoterpene oxide and it has several biological effects including hepatoprotective effects. In this study we revealed that 1,8-cineole ameliorates NASH of Pten KO mice. Pten KO mice were assigned to a control group without any medication or to a 1,8-cineole group injected with 50 mg/kg i.p. twice per week for eight weeks. At eight weeks, livers from each group were processed to measure triglyceride (TG) content, gene expression analysis, western blot analysis, and histological examination including Oil red O staining. 1,8-cineole ameliorated hepatic steatosis in Pten KO mice, revealed by TG content and Oil red O staining. Moreover, 1,8-cineole downregulated collagen 1a1 expression and improved liver fibrosis. Thus, 1,8-cineole has potential as a candidate to treat NASH by inactivating the Akt/PI3-kinase pathway.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glukose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glukose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
Eucalyptol, 99%
Sigma-Aldrich
D-(+)-Glukose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glukose, ACS reagent
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
DL-Glyceraldehyd-3-phosphat -Lösung, 45-55 mg/mL in H2O
Sigma-Aldrich
D-(+)-Glukose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
D-(+)-Glukose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glukose, 99.9 atom % 16O, 99.9 atom % 12C
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)