Merck
  • Molecular cloning and characterization of the gonadotropin subunits GPα, FSHβ, and LHβ genes in the stinging catfish Heteropneustes fossilis: phylogeny, seasonal expression and pituitary localization.
Molecular cloning and characterization of the gonadotropin subunits GPα, FSHβ, and LHβ genes in the stinging catfish Heteropneustes fossilis: phylogeny, seasonal expression and pituitary localization.
Journal of experimental zoology. Part A, Ecological genetics and physiology (2015-07-25)
Arup Acharjee, Radha Chaube, Keerikkattil Paily Joy
ZUSAMMENFASSUNG
Gonadotropins are heterodimeric glycoproteins secreted by the pituitary, and consist of a common glycoprotein hormone alpha (GPα) and the function-specific follicle-stimulating hormone beta subunit (FSHβ) or luteinizing hormone beta subunit (LHβ). In the present study, the subunit protein genes were cloned and characterized from the pituitary of the catfish Heteropneustes fossilis. Full-length cDNAs of GPα, FSHβ, and LHβ are 511 base pairs (bp), 659 bp and 660 bp long, and encode 92, 108, and 112 aminoacids long mature proteins, respectively. GPα has 10 cysteines with 2 N-linked glycosylation sites while LHβ contains 12 cysteines with a single N-linked glycosylation site. In contrast, FSHβ has 13 cysteines, 1 additional over the conserved 12 cysteines of other vertebrates, and a single glycosylation site between Cys 3 and Cys 4. Phylogenetic analyses of the deduced proteins confirm their homology and relationships with the respective gonadotropin subunit proteins of gnathostome vertebrates. Tissue expression analysis by semi-quantitative RT-PCR shows that GPα mRNA is expressed only in the pituitary while both FSHβ and LHβ mRNA are expressed in extra-pituitary sites. The subunit mRNAs show both seasonal and sex dimorphic variations especially in the expression of FSHβ and LHβ transcripts. In the sexually quiescent phase, the transcript expression is low while in the recrudescent phase, the expressions are differential, high, and varied with regard to sex and reproductive phase. In situ hybridization of the mRNAs gave positive signals in gonadotropes in the pars distalis of the pituitary, which exhibited seasonal variation in staining intensity and numbers.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Trizma® Base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® Base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, suitable for electrophoresis, Molecular Biology, 99.0-101.0% (titration)
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, ACS reagent, 99.0-101.0%
Sigma-Aldrich
Tris(hydroxymethyl)aminomethan, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamin, meets USP testing specifications
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Trizma® Base, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz -Lösung, Molecular Biology, 0.5 M in H2O, DNase, RNase, NICKase and protease, none detected
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Ethyl-3-aminobenzoat -methansulfonat, 98%
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, reagent grade, 98.5-101.5% (titration)
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Trizma® Base, ≥99.0% (T)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Trizma® Base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, Sigma Grade, suitable for plant cell culture, 98.5-101.5%
Sigma-Aldrich
Trizma® Base, ≥99.9% (titration), crystalline
Sigma-Aldrich
Ethylendiamintetraessigsäure Dinatriumsalz Dihydrat, meets analytical specification of Ph. Eur., BP, USP, FCC, 99.0-101.0%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
o-Xylen, anhydrous, 97%