Merck
  • Influence of intraspecific variability and abiotic factors on mycotoxin production in Penicillium roqueforti.
Influence of intraspecific variability and abiotic factors on mycotoxin production in Penicillium roqueforti.
International journal of food microbiology (2015-09-01)
Kévin Fontaine, Nolwenn Hymery, Marlène Z Lacroix, Sylvie Puel, Olivier Puel, Karim Rigalma, Vincent Gaydou, Emmanuel Coton, Jérôme Mounier
ZUSAMMENFASSUNG
Penicillium roqueforti has the ability to produce secondary metabolites, including roquefortine C (ROQC) and mycophenolic acid (MPA). In a previous study, the presence of these mycotoxins, alone or in co-occurrence, has been reported in blue-veined cheese. A high variability of mycotoxin content has also been observed, although the majority of samples exhibited relatively low concentrations. The observed variability raises the question of the factors impacting ROQC and MPA production. In this context, the mycotoxigenic potential of 96 P. roqueforti strains (biotic factor) and the effect of some abiotic factors (pH, temperature, NaCl and O2 contents, and C/N ratio) on mycotoxin production were evaluated. A high intraspecific diversity, established via genotypic (RAPD) and phenotypic (FTIR) approaches, was observed. It was associated with mycotoxigenic potential variability and may thus explain part of the observed variability in mycotoxin content of blue-veined cheese. Moreover, a significant decrease of ROQC and MPA production was observed for conditions (temperature, C/N ratio, O2 and NaCl concentrations) encountered during cheese-making compared with optimal growth conditions. The results also highlighted that there was no significant effect of addition of ROQC amino-acid precursor on the production of both mycotoxins whereas a pH increase from 4.5 to 6.5 slightly reduced MPA but not ROQC production.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
1-Methoxy-2-propyl-acetat, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Milchsäure, meets USP testing specifications
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Milchsäure, 88%, FCC
Sigma-Aldrich
Milchsäure, natural, ≥85%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Milchsäure -Lösung, ACS reagent, ≥85%
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
DL-Milchsäure, 85 % (w/w), syrup
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis