Merck
Outer-membrane translocation of bulky small molecules by passive diffusion.
Proceedings of the National Academy of Sciences of the United States of America (2015-05-28)
Bert van den Berg, Satya Prathyusha Bhamidimarri, Jigneshkumar Dahyabhai Prajapati, Ulrich Kleinekathöfer, Mathias Winterhalter
ZUSAMMENFASSUNG
The outer membrane (OM) of gram-negative bacteria forms a protective layer around the cell that serves as a permeability barrier to prevent unrestricted access of noxious substances. The permeability barrier of the OM results partly from the limited pore diameters of OM diffusion channels. As a consequence, there is an "OM size-exclusion limit," and the uptake of bulky molecules with molecular masses of more than ∼ 600 Da is thought to be mediated by TonB-dependent, active transporters. Intriguingly, the OM protein CymA from Klebsiella oxytoca does not depend on TonB but nevertheless mediates efficient OM passage of cyclodextrins with diameters of up to ∼ 15 Å. Here we show, by using X-ray crystallography, molecular dynamics simulations, and single-channel electrophysiology, that CymA forms a monomeric 14-stranded β-barrel with a large pore that is occluded on the periplasmic side by the N-terminal 15 residues of the protein. Representing a previously unidentified paradigm in OM transport, CymA mediates the passive diffusion of bulky molecules via an elegant transport mechanism in which a mobile element formed by the N terminus acts as a ligand-expelled gate to preserve the permeability barrier of the OM.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid -Lösung, ~30% in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumphosphat, tribasisch, reagent grade, ≥97%
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
N,N-Dimethyldodecylamin-N-oxid, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, ≥98% (HPLC), powder
Sigma-Aldrich
L-Thyroxin Natriumsalz Pentahydrat, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Spautin-1, ≥98% (HPLC)