Merck
  • Enrichment of nuclear S100A4 during G2/M in colorectal cancer cells: possible association with cyclin B1 and centrosomes.
Enrichment of nuclear S100A4 during G2/M in colorectal cancer cells: possible association with cyclin B1 and centrosomes.
Clinical & experimental metastasis (2015-09-10)
Eivind Valen Egeland, Kjetil Boye, Solveig J Pettersen, Mads H Haugen, Tove Øyjord, Lene Malerød, Kjersti Flatmark, Gunhild M Mælandsmo
ZUSAMMENFASSUNG
S100A4 promotes metastasis in several types of cancer, but the involved molecular mechanisms are still incompletely described. The protein is associated with a wide variety of biological functions and it locates to different subcellular compartments, including nuclei, cytoplasm and extracellular space. Nuclear expression of S100A4 has been associated with more advanced disease stage as well as poor outcome in colorectal cancer (CRC). The present study was initiated to investigate the nuclear function of S100A4 and thereby unravel potential biological mechanisms linking nuclear expression to a more aggressive phenotype. CRC cell lines show heterogeneity in nuclear S100A4 expression and preliminary experiments revealed cells in G2/M to have increased nuclear accumulation compared to G1 and S cells, respectively. Synchronization experiments validated nuclear S100A4 expression to be most prominent in the G2/M phase, but manipulating nuclear levels of S100A4 using lentiviral modified cells failed to induce changes in cell cycle distribution and proliferation. Proximity ligation assay did, however, demonstrate proximity between S100A4 and cyclin B1 in vitro, while confocal microscopy showed S100A4 to localize to areas corresponding to centrosomes in mitotic cells prior to chromosome segregation. This might indicate a novel and uncharacterized function of the metastasis-associated protein in CRC cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, Molecular Biology
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Kaliumchlorid, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Kaliumchlorid, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Kaliumchlorid, Molecular Biology, ≥99.0%
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Kaliumchlorid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99.0%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Kaliumchlorid, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E508, 99-100.5% (AT), ≤0.0001% Al
Sigma-Aldrich
Thymidin, powder, BioReagent, suitable for cell culture