Merck
  • Familial hypercholesterolemia mutations in the Middle Eastern and North African region: a need for a national registry.
Familial hypercholesterolemia mutations in the Middle Eastern and North African region: a need for a national registry.
Journal of clinical lipidology (2015-04-26)
Mary Aderayo Bamimore, Ahmed Zaid, Yajnavalka Banerjee, Ahmad Al-Sarraf, Marianne Abifadel, Nabil G Seidah, Khalid Al-Waili, Khalid Al-Rasadi, Zuhier Awan
ZUSAMMENFASSUNG
Familial hypercholesterolemia (FH) is a well-understood Mendelian disorder that increases the risk of cardiovascular disease (CVD), a leading cause of mortality in Middle Eastern and North African (MENA) countries. Review the reporting status of FH mutations across MENA and propose a systemic and strategic method for building a MENA FH registry. Systematic literature search for statistics pertaining to CVD and comparison of number of FH mutations reported in MENA countries and countries with established FH registries. Only 57 mutations were reported in 17 MENA countries, whereas more than 500 mutations reported in 3 Western countries. Mortality rates due to CVD were significantly higher in MENA countries compared with Western countries. The relatively low reporting of FH mutations in the consanguineous MENA communities with higher prevalence of CVD indicates poor awareness of CVD genetic risk and warrants a registry to prevent premature CVD due to FH. This registry will help in identifying novel and reported FH mutations, all of which will have clinical and research benefits in MENA countries.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cholesterin, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterin, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
SyntheChol® NS0-Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Cholesterin, from sheep wool, ≥92.5% (GC), powder
SAFC
Cholesterin, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Cholesterin, from lanolin, ≥99.0% (GC)