Merck
  • RNA-sequencing reveals transcriptional up-regulation of Trem2 in response to bexarotene treatment.
RNA-sequencing reveals transcriptional up-regulation of Trem2 in response to bexarotene treatment.
Neurobiology of disease (2015-06-15)
Iliya Lefterov, Jonathan Schug, Anais Mounier, Kyong Nyon Nam, Nicholas F Fitz, Radosveta Koldamova
ZUSAMMENFASSUNG
We have recently demonstrated that short term bexarotene treatment of APP/PS1 mice significantly improves their cognitive performance. While there were no changes in plaque load, or insoluble Aβ levels in brain, biochemical analysis strongly suggested improved clearance of soluble Aβ, including Aβ oligomers. To get further insight into molecular mechanisms underlying this therapeutic effect, we explored genome-wide differential gene expression in brain of bexarotene and control treated APP/PS1 mice. We performed high throughput massively parallel sequencing on mRNA libraries generated from cortices of bexarotene or vehicle treated APP/PS1 mice and compared the expression profiles for differential gene expression. Gene Ontology (GO) Biological Process categories with the highest fold enrichment and lowest False Discovery Rate (FDR) are clustered in GO terms immune response, inflammatory response, oxidation-reduction and immunoglobulin mediated immune response. Chromatin immunoprecipitation (ChIP) followed by ChIP-QPCR, and RT-QPCR expression assays were used to validate select genes, including Trem2, Tyrobp, Apoe and Ttr, differentially expressed in response to Retinoid X Receptor (RXR) activation. We found that bexarotene significantly increased the phagocytosis of soluble and insoluble Aβ in BV2 cells. The results of our study demonstrate that in AD model mice expressing human APP, gene networks up-regulated in response to RXR activation by the specific, small molecule, ligand bexarotene may influence diverse regulatory pathways that are considered critical for cognitive performance, inflammatory response and Aβ clearance, and may provide an explanation of the bexarotene therapeutic effect at the molecular level. This study also confirms that unbiased massive parallel sequencing approaches are useful and highly informative for revealing brain molecular and cellular mechanisms underlying responses to activated nuclear hormone receptors in AD animal models.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Glycerin, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerin, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerin, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
1,1,1,3,3,3-Hexafluor-2-propanol, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyd -Lösung, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehyd -Lösung, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Glycerin, ≥99.5%
Sigma-Aldrich
Glycerin -Lösung, 83.5-89.5% (T)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Glycerin, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Glycerin, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Sigma-Aldrich
Formaldehyd -Lösung, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Glycerin, BioXtra, ≥99% (GC)
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Glycerin, FCC, FG
Sigma-Aldrich
Formaldehyd -Lösung, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Supelco
Formaldehyd -Lösung, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Dimethylsulfoxid -Lösung, 50 wt. % in H2O
Sigma-Aldrich
Bexarotene, ≥98% (HPLC)
Sigma-Aldrich
Formaldehyd-12C -Lösung, 20% in H2O, 99.9 atom % 12C