Merck
  • Functionalization of electrospun poly(caprolactone) fibers for pH-controlled delivery of doxorubicin hydrochloride.
Functionalization of electrospun poly(caprolactone) fibers for pH-controlled delivery of doxorubicin hydrochloride.
Journal of biomaterials science. Polymer edition (2015-09-26)
Manisha Jassal, Sukalyan Sengupta, Sankha Bhowmick
ZUSAMMENFASSUNG
Functionalized electrospun polymer fibers are a promising candidate for controlled delivery of chemotherapeutic drugs to improve the therapeutic efficacy and to reduce the potential toxic effects by delivering the drug at a rate governed by the physiological need of the site of action. In this study, poly(caprolactone) (PCL) fibers were fabricated by electrospinning, followed by hydrolyzation to introduce functional groups on the fiber surface. Characterization studies were performed on these functionalized fibers using X-ray photoelectron spectroscopy, scanning electron microscopy, and Toluidine Blue O dye assay. The pH-sensitivity of the functional groups on the fiber surface and doxorubicin hydrochloride was utilized to bind the drug electrostatically to these functionalized PCL fibers. The effect of pH on drug loading and release kinetics was investigated. Results indicate successful electrostatic binding of the drug to functionalized electrospun fibers and a high drug payload. The drug delivery response can be modulated by introduction of suitable stimuli (pH).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethylformamid, ACS reagent, ≥99.8%
Sigma-Aldrich
Dichlormethan, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
N,N-Dimethylformamid, anhydrous, 99.8%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
N,N-Dimethylformamid, ReagentPlus®, ≥99%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis
Sigma-Aldrich
N,N-Dimethylformamid, Molecular Biology, ≥99%
Sigma-Aldrich
Natronlauge, 5.0 M
Sigma-Aldrich
Natriumhydroxid, beads, 16-60 mesh, reagent grade, 97%
Sigma-Aldrich
N,N-Dimethylformamid, biotech. grade, ≥99.9%
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, flakes
Sigma-Aldrich
Natriumhydroxid, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Natriumhydroxid, ultra dry, powder or crystals, 99.99% trace metals basis
Sigma-Aldrich
Dichlormethan, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichlormethan, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Natriumhydroxid, anhydrous, free-flowing, Redi-Dri, reagent grade, ≥98%, pellets
Sigma-Aldrich
3-Ethyl-2,4-pentandion, Tautormermischung, 98%
Sigma-Aldrich
Natriumhydroxid-16O -Lösung, 20 wt. % in H216O, 99.9 atom % 16O