Merck
Effect of ultrasonic waves on the stability of all-trans lutein and its degradation kinetics.
Ultrasonics sonochemistry (2015-05-03)
Jiang-Feng Song, Da-Jing Li, Hui-Li Pang, Chun-Quan Liu
ZUSAMMENFASSUNG
Ultrasound treatment has been widely applied in the extraction of biologically active compounds including carotenoids. However, there are few reports on their effects on the stability of these compounds. In the present study, the stability of all-trans lutein, one of the carotenoids, was investigated under the action of ultrasound. Results showed that ultrasound induced the isomerization of all-trans lutein to its isomers, namely to 13-cis lutein, 13'-cis lutein, 9-cis lutein and 9'-cis lutein as analyzed by HPLC coupled with DAD and LC-MS; and the percentage of the isomerization increased with increasing both ultrasonic frequency and power. The stability of all-trans lutein in dichloromethane was worst among multiple kinds of solvents. Interestingly, the retention rate of all-trans lutein improved as the temperature increased, which runs counter to the Arrhenius law. Under ultrasound irradiation, the degradation mechanism might be different with various temperatures, the degradation of all-trans lutein followed first-order kinetics at 20°C, while second-order kinetics was followed at 30-50°C. As the ultrasonic reaction time prolonged, lutein epoxidation nearly occurred. Those results presented here emphasized that UAE techniques should be carefully used in the extraction of all-trans lutein.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
Sigma-Aldrich
Dichlormethan, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Tetrahydrofuran, anhydrous, contains 250 ppm BHT as inhibitor, ≥99.9%
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethylacetat, anhydrous, 99.8%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Stickstoff, ≥99.998%
Sigma-Aldrich
Ethylacetat, ≥99%, FCC, FG
Sigma-Aldrich
Aceton, ≥99%, FCC, FG
Supelco
Tetrahydrofuran, HPLC grade, ≥99.9%, inhibitor-free
Sigma-Aldrich
Ethylacetat
Sigma-Aldrich
Dichlormethan, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Xanthophyll, from marigold
Sigma-Aldrich
Aceton, natural, ≥97%
Sigma-Aldrich
Aceton, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Ethylacetat, natural, ≥99%, FCC, FG
Sigma-Aldrich
Ethylacetat, ReagentPlus®, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Sigma-Aldrich
LDN-57444, ≥98% (HPLC)