Merck
  • Recombinant Human Semenogelin-1 (Sg1) and Sg1 (1-159) form Detergent Stable Amyloid like Aggregates in vitro.
Recombinant Human Semenogelin-1 (Sg1) and Sg1 (1-159) form Detergent Stable Amyloid like Aggregates in vitro.
Protein and peptide letters (2015-11-10)
Neetu Sharma, S Vishwanath, Basant K Patel
ZUSAMMENFASSUNG
Senile seminal vesicle amyloidosis (SSVA) is associated with deposition of semenogelin-1 (Sg1) protein aggregates in seminal vesicles that may manifest as hematospermia. Sg1 is the predominant protein that entraps spermatozoa which are freed upon fragmentation of Sg1 by the protease prostate specific antigen (PSA), post semen release. Certain small peptide fragments of Sg1 have been reported to form amyloid aggregates in vitro that can enhance HIV infectivity to cell cultures. However, the amyloid deposits in the seminal vesicles are expected to be that of the full length Sg1, as PSA is encountered downstream. So far, amyloid forming ability of full length Sg1 has not been established in vitro. Here, we examined the amyloidogenicity of full length Sg1 and a large fragment Sg1 (1-159), using recombinant proteins and tested if Zinc has any effect on their aggregation. Levels of Zinc, which is essential for health of male reproductive system, gradually decline with age. We succeeded in forming amyloid-like aggregates of Sg1 full length and Sg1 (1-159) fragment showing detergent stability and found that presence of Zn2+ substantially inhibits their amyloid aggregation in vitro. Possibly, high Zn2+ found in seminal plasma of young individuals may have preventive role against aggregation of Sg1 in seminal vesicles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumdodecylsulfat, BioReagent, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Guanidin -hydrochlorid, Molecular Biology, ≥99%
Sigma-Aldrich
Imidazol, ReagentPlus®, 99%
Sigma-Aldrich
Natriumdodecylsulfat, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Imidazol, ACS reagent, ≥99% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ACS reagent, ≥99.0%
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, Molecular Biology, 10% in H2O
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Guanidin -hydrochlorid, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Natriumdodecylsulfat, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.4-100.6%, powder
Sigma-Aldrich
Guanidin -hydrochlorid, ≥98%
Sigma-Aldrich
Imidazol, Molecular Biology, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumdodecylsulfat, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Natriumdodecylsulfat -Lösung, BioUltra, 20% in H2O
Sigma-Aldrich
Ampicillin, anhydrous, 96.0-102.0% (anhydrous basis)
Sigma-Aldrich
Imidazol, BioUltra, ≥99.5% (GC)
Supelco
Natriumdodecylsulfat, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Imidazol-Pufferlösung, BioUltra, 1 M in H2O
Sigma-Aldrich
Imidazol, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Natriumdodecylsulfat, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Guanidin -hydrochlorid -Lösung, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Natriumdodecylsulfat, ≥98.0% (GC)
Sigma-Aldrich
3-Aminophenol, 98%