Merck
  • A new (68)Ga anionic concentration and purification method for automated synthesis of [(68)Ga]-DOTA or NODAGA conjugated peptides in high radiochemical purity.
A new (68)Ga anionic concentration and purification method for automated synthesis of [(68)Ga]-DOTA or NODAGA conjugated peptides in high radiochemical purity.
Journal of labelled compounds & radiopharmaceuticals (2015-08-06)
Rana Ben Azzouna, Faisal Alshoukr, Sébastien Leygnac, Alexandre Guez, Walter Gonzalez, Olivier Rousseaux, Denis Guilloteau, Dominique Le Guludec
ZUSAMMENFASSUNG
The (68)Ge/(68)Ga generator is of increasing interest for clinical PET. For successful labelling, the eluate has to be purified. The aim of our approach is to improve the existing anionic methods which have a number of advantages compared to other methods but which use high concentrated HCl, and require an additional anionizing step. A new (68)Ga-eluate anionic purification method that enables rapid and high efficiency labelling of DOTA and NODAGA conjugated peptides in high radiochemical purity is described. The new method uses NaCl as an alternative Cl(-) source to the corrosive HCl and combines the three standard steps in a single step. The recovery yield was ≥90%, and the (68)Ge breakthrough was in conformity with the European Pharmacopeia limit. An automated labelling of DOTA and NODAGA-conjugated peptides was performed with the new method, using acetate sodium buffer, with a total duration of 13 min and a radiochemical yield >85%. The labelled peptides have a radiochemical purity exceeding 99% and can be used directly without any further purification step and without the quality control by gas chromatography. Furthermore, the new method has an economic advantage: it offers the possibility to use generator until 20 months after the calibration date.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Supelco
Salzsäure -Lösung, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Natriumacetat, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
L-Lysin -monohydrochlorid, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumacetat, 99.995% trace metals basis
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Salzsäure -Lösung, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Natriumacetat, >99%, FG
Sigma-Aldrich
Chlorwasserstoff, ReagentPlus®, ≥99%
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Natriumacetat, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%