Merck
Assembly and maintenance of the flagellum attachment zone filament in Trypanosoma brucei.
Journal of cell science (2015-05-15)
Qing Zhou, Huiqing Hu, Cynthia Y He, Ziyin Li
ZUSAMMENFASSUNG
Adhesion of motile flagella to the cell body in Trypanosoma brucei requires a filamentous cytoskeletal structure termed the flagellum attachment zone (FAZ). Despite its essentiality, the complete molecular composition of the FAZ filament and its roles in FAZ filament assembly remain poorly understood. By localization-based screening, we here identified a new FAZ protein, which we called FAZ2. Knockdown of FAZ2 disrupted the FAZ filament, destabilized multiple FAZ filament proteins and caused a cytokinesis defect. We also showed that FAZ2 depletion destabilized another new FAZ filament protein and several flagellum and cytoskeleton proteins. Furthermore, we identified CC2D and KMP11 as FAZ2 partners through affinity purification, and showed that they are each required for maintaining a stable complex. Finally, we demonstrated that FAZ filament proteins are incorporated into the FAZ filament from the proximal region, in contrast to the flagellum components, which are incorporated from the distal tip. In summary, we identified three new FAZ filament proteins and a FAZ filament protein complex, and our results suggest that assembly of the FAZ filament occurs at the proximal region and is essential to maintain the stability of FAZ filament proteins.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Harnstoff, meets USP testing specifications
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat Hydrat, 98%
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Harnstoff -Lösung, 40 % (w/v) in H2O
Sigma-Aldrich
Harnstoff-12C, 99.9 atom % 12C