Merck
  • Tackling proteome changes in the longissimus thoracis bovine muscle in response to pre-slaughter stress.
Tackling proteome changes in the longissimus thoracis bovine muscle in response to pre-slaughter stress.
Journal of proteomics (2015-04-11)
Daniel Franco, Ariadna Mato, Francisco J Salgado, María López-Pedrouso, Mónica Carrera, Susana Bravo, María Parrado, José M Gallardo, Carlos Zapata
ZUSAMMENFASSUNG
Pre-slaughter stress has adverse effects on meat quality that can lead to the occurrence of Dark Firm Dry (DFD) meat in cattle. This study explores the previously uncharacterized proteome changes linked to pre-slaughter stress in the longissimus thoracis (LT) bovine muscle. Differential proteome profiles of DFD and normal (non-DFD) LT meat samples from male calves of the Rubia Gallega breed were assessed by 2-DE coupled to MS analysis (LC-MS/MS and MALDI TOF/TOF MS). A total of seven structural-contractile proteins (three different myosin light chain isoforms, two fast skeletal myosin light chain 2 isoforms, troponin C type 2 and cofilin-2) and three metabolism enzymes (triosephosphate isomerase, ATP synthase and beta-galactoside alpha-2,6-sialyltransferase) were found to have statistically significant differential abundance in sample groups. In addition, 2-DE in combination with the phosphoprotein-specific fluorescent dye Pro-Q DPS revealed that highly phosphorylated fast skeletal myosin regulatory light chain 2 isoforms underwent the most intense relative change in muscle conversion to DFD meat. Therefore, they appear to be the most sensitive biomarkers of stress just prior to slaughter in Rubia Gallega. Overall, these findings will facilitate a more integrative understanding of the biochemical processes associated with stress in cattle muscle and their effects in meat quality. Pre-slaughter stress is a crucial factor in meat production. Animals destined for slaughter are stressed by a variety of endogenous and exogenous factors that negatively affect the complex post-mortem biochemical events underlying the conversion of muscle into meat. The study of the muscle proteome has a great relevance for understanding the molecular mechanisms associated with stress. However, there is no information available on the molecular changes linked to pre-slaughter stress in cattle on the proteome scale. Our study led to the identification of a number of candidate proteins associated with the response to pre-slaughter stress in the LT bovine muscle of the Rubia Gallega breed. The functions of those significantly changed proteins have a clear biological relationship with stress response. These findings contribute to a deeper insight into the molecular pathways that respond to stress in cattle.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Acetonitril, anhydrous, 99.8%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Jodacetamid, BioUltra
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Jodacetamid, Single use vial of 56 mg
Sigma-Aldrich
Harnstoff, powder, BioReagent, Molecular Biology, suitable for cell culture
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Jodacetamid, ≥99% (NMR), crystalline
Sigma-Aldrich
Harnstoff, ACS reagent, 99.0-100.5%
Supelco
Harnstoff, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Harnstoff -Lösung, BioUltra, ~8 M in H2O
Sigma-Aldrich
Thioharnstoff, ACS reagent, ≥99.0%
Sigma-Aldrich
Harnstoff, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Harnstoff, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Harnstoff, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Sigma-Aldrich
Thioharnstoff, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Essigsäure, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetonitril, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
5α-Androstan-17β-ol-3-on, ≥97.5%
Sigma-Aldrich
Harnstoff, suitable for electrophoresis
Sigma-Aldrich
DL-Methionin, ≥99%
Sigma-Aldrich
Harnstoff, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Essigsäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Essigsäure, natural, ≥99.5%, FG
Sigma-Aldrich
Harnstoff, meets USP testing specifications
SAFC
Jodacetamid
Sigma-Aldrich
3-[(3-Cholamidopropyl)dimethylammonio]-1-propansulfonat Hydrat, 98%
Sigma-Aldrich
Harnstoff, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
DL-Methionin, 99%, FCC, FG
Sigma-Aldrich
DL-Cystein, technical grade