Merck
Nonomuraea indica sp. nov., novel actinomycetes isolated from lime-stone open pit mine, India.
The Journal of antibiotics (2015-03-19)
Syed Raziuddin Quadri, Xin-Peng Tian, Jing Zhang, Jie Li, Guo-Xing Nie, Shu-Kun Tang, Jamal Al Ruwaili, Dayanand Agsar, Wen-Jun Li, Syed G Dastager
ZUSAMMENFASSUNG
A Gram-positive, aerobic, nonmotile actinomycete strain designated DRQ-2(T) was isolated from the soil sample collected from lime-stone open pit mine from the Gulbarga region, Karnataka province, India. Strain DRQ-2(T) was identified as a member of the genus Nonomuraea by a polyphasic approach. Strain DRQ-2(T) could be differentiated from other members of the genus Nonomuraea on the basis of physiology and 16S rRNA gene sequence analysis. The 16S rRNA gene sequence similarity of strain DRQ-2(T) showed highest sequence similarity to Nonomuraea muscovyensis DSM 45913(T) (99.1%), N. salmonea DSM 43678(T) (98.2%) and N. maheshkhaliensis JCM 13929(T) with 98.0%, respectively. Chemotaxonomic properties showing predominant menaquinones of MK-9 (H4), MK-9(H2) and MK-9(H6), major polar lipids comprised diphosphatidylglycerol, phosphatidylmono methyl ethanolamine (PME), phosphatidylethanolamine (PE), hydroxy-PME (OH-PME), hydroxy PE (OH-PEE), phosphatidylglycerol (PG), ninhydrin-positive phosphoglycolipid and unknown phospholipid, fatty acids with major amounts of i-C16:0, ai-C15:0 and ai-C17:0 supported allocation of the strain to the genus Nonomuraea. Results of DNA-DNA hybridization and physiological tests allowed genotypic and phenotypic differentiation of strain DRQ-2(T) from closely related species. The genomic DNA G+C content of the organism was 72.5 mol%. On the basis of phenotypic, chemotypic and molecular characteristics, strain DRQ-2(T) represents a novel species of the genus Nonomuraea, for which the name N. indica sp. nov. is proposed, with type strain DRQ-2(T) (=NCIM 5480(T)= CCTCC AA 209050(T)).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumphosphat, 96%
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M
SAFC
Natriumchlorid -Lösung, 5 M
Sigma-Aldrich
Natriumchlorid, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Natriumchlorid -Lösung, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Natriumchlorid, 99.999% trace metals basis
Sigma-Aldrich
Natriumchlorid, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 0.85%
Sigma-Aldrich
Natriumchlorid, tablet
Sigma-Aldrich
Natriumchlorid-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Natriumchlorid, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Natriumchlorid, AnhydroBeads, −10 mesh, 99.999% trace metals basis