Merck
  • Regulated C-C motif ligand 2 (CCL2) in luteal cells contributes to macrophage infiltration into the human corpus luteum during luteolysis.
Regulated C-C motif ligand 2 (CCL2) in luteal cells contributes to macrophage infiltration into the human corpus luteum during luteolysis.
Molecular human reproduction (2015-05-25)
Junko Nio-Kobayashi, Masataka Kudo, Noriaki Sakuragi, Shunsuke Kimura, Toshihiko Iwanaga, W Colin Duncan
ZUSAMMENFASSUNG
Intense macrophage infiltration is observed during luteolysis in various animals including women; however, we still do not know how macrophage infiltration into the human corpus luteum (CL) during luteolysis is regulated. In this study, we examined the expression, localization and regulation of an important chemokine for the recruitment of monocyte/macrophage lineages, C-C motif ligand 2 (CCL2), in the human CL across the luteal phase and in cultured human luteinized granulosa cells (LGCs), with special reference to the number of infiltrating macrophages and luteal cell function. CCL2 mRNA increased in the non-functional regressing CL during menstruation (P < 0.01), corresponding to an elevated mRNA expression of a macrophage-derived cytokine, tumor necrosis factor (TNF), and an increased number of infiltrating macrophages positively stained with a macrophage marker, CD68. CCL2 protein was immunohistochemically localized to the cytoplasm of granulosa-lutein and theca-lutein cells, and CCL2 mRNA was significantly reduced by hCG both in vivo (P < 0.05) and in vitro (P < 0.01). CCL2 was also down-regulated by luteotrophic prostaglandin (PG) E (P < 0.0001), but up-regulated by luteolytic PGF (P < 0.05) in vitro. Administration of TNF significantly enhanced the CCL2 mRNA expression in cultured LGCs (P < 0.01). A greater abundance of infiltrating macrophages were found around granulosa-lutein cells lacking 3β-HSD or PGE synthase (PGES) immunostaining. CCL2 mRNA expression was negatively correlated with both HSD3B1 and PGES, suggesting that locally produced progesterone and PGE suppress macrophage infiltration into the CL. Taken together, the infiltration of macrophages in the human CL is regulated by endocrine and paracrine molecules via regulation of the CCL2 expression in luteal cells.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Wasser, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Wasser, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, ≥99% (TLC), film or powder
Sigma-Aldrich
PMA, for use in molecular biology applications, ≥99% (HPLC), Molecular Biology
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Wasser, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Selen, powder, −100 mesh, 99.99% trace metals basis
Sigma-Aldrich
Wasser, Molecular Biology, sterile filtered
SAFC
L-Glutamin
Sigma-Aldrich
Fluorescein, free acid
Sigma-Aldrich
Selen, powder, −100 mesh, ≥99.5% trace metals basis
Sigma-Aldrich
Wasser, BioPerformance Certified
Sigma-Aldrich
Zyklisches Adenosin-3′,5′-Monophosphat, ≥98.5% (HPLC), powder
Sigma-Aldrich
Phorbol-12-myristat-13-acetat, synthetic, ≥98.0% (TLC)
Sigma-Aldrich
Wasserstoffperoxid -Lösung, 34.5-36.5%
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Wasser, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Wasser, PCR Reagent
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
Selen, pellets, <5 mm particle size, ≥99.999% trace metals basis
Sigma-Aldrich
Selen, pellets, <5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Wasser, endotoxin, free
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Selen, foil, 25x25mm, thickness 3mm, 99.95%
Sigma-Aldrich
Wasser-16O, ≥99.94 atom % 16O
Selen, pellets, < 5mm, ≥99.999%