Merck
  • Activation of mitochondrial STAT-3 and reduced mitochondria damage during hypothermia treatment for post-cardiac arrest myocardial dysfunction.
Activation of mitochondrial STAT-3 and reduced mitochondria damage during hypothermia treatment for post-cardiac arrest myocardial dysfunction.
Basic research in cardiology (2015-10-17)
Chien-Hua Huang, Min-Shan Tsai, Chih-Yen Chiang, Yu-Jen Su, Tzung-Dau Wang, Wei-Tien Chang, Huei-Wen Chen, Wen-Jone Chen
ZUSAMMENFASSUNG
While therapeutic hypothermia improves the outcomes of individuals in cardiac arrest, the hemodynamic responses and mechanisms which underlie hypothermia-induced cardioprotection are not fully understood. Therefore, we investigated the mechanism by which induced hypothermia preserves cardiac function and protects against mitochondrial damage following cardiac arrest. Cardiac arrest was induced in adult male Wistar rats by asphyxiation for 8.5 min. Following resuscitation, the animals were randomly assigned to a hypothermia (32 °C) or normothermia (37 °C) group. Monitoring results showed that cardiac output at the fourth hour after resuscitation was significantly better in rats treated with hypothermia when compared to rats treated with normothermia (P < 0.01). Examinations by transmission electron microscopy showed that mitochondria in the left ventricle of rats in the hypothermia group were significantly less swollen compared to such mitochondria in the normothermia group (P < 0.001). Additionally, opening of mitochondrial permeability transition pores occurred less frequently in the hypothermic group. While complex I/III activity in the electron transport reaction was damaged after cardiac arrest and resuscitation, the degree of injury was ameliorated by hypothermia treatment (P < 0.05). The amount of STAT-3 phosphorylated at tyrosine 705 and its expression in mitochondria were significantly higher under hypothermia treatment compared to normothermia treatment. In vitro studies showed that inhibition STAT-3 activation abolished the ability of hypothermia to protect H9C2 cardiomyocytes against injury produced by simulated ischemia and reperfusion. Therapeutic hypothermia treatment can ameliorate cardiac dysfunction and help preserve both mitochondrial integrity and electron transport activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Saccharose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Magnesiumchlorid -Lösung, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Magnesiumchlorid, ≥98%
Sigma-Aldrich
Saccharose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC)
Sigma-Aldrich
Saccharose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesiumchlorid, powder, <200 μm
Sigma-Aldrich
Saccharose, ACS reagent
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
Saccharose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesiumchlorid, suitable for insect cell culture, BioReagent, ≥97.0%
Sigma-Aldrich
Saccharose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Magnesiumchlorid -Lösung, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Saccharose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Saccharose, meets USP testing specifications
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.9% trace metals basis
Sigma-Aldrich
Magnesiumchlorid, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
DL-Serin, ≥98% (TLC)
Sigma-Aldrich
Magnesiumchlorid -Lösung, 0.1 M
Sigma-Aldrich
DL-Serin, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98% (HPLC)
Sigma-Aldrich
DL-Tyrosin, 99%
Sigma-Aldrich
Magnesiumchlorid -Lösung, BioUltra, Molecular Biology, ~0.025 M in H2O