Merck
Biological removal of triphenylmethane dyes from aqueous solution by Lemna minor.
Acta chimica Slovenica (2015-06-19)
Anamaria Török, Erzsébet Buta, Cerasella Indolean, Szende Tonk, Luminita Silaghi-Dumitrescu, Cornelia Majdik
ZUSAMMENFASSUNG
The aim of this study is to investigate and develop a phytoremediation method for the removal of two triphenylmethane dyes (crystal violet and malachite green) using an aquatic plant, Lemna minor. The effects of operational parameters such as aquatic plant quantity, initial dye concentration, initial pH of the solutions and temperature of the medium were studied in order to determine the optimum phytoremediation conditions. The plant's photosynthetic pigments were determined quantitatively in order to detect the plant's response to abiotic stress. During the phytoremediation experiments the parallel sub-processes (phytosorption, phytoextraction, phytodegradation) were observed and analysed. The mechanisms of phytoremediation were studied using Fourier transformation infrared spectroscopy, ultraviolet-visible spectroscopy, thin layer chromatography and Energy-dispersive X-ray spectroscopy. Results show that the plant tolerated high concentrations (300 mg/L) of dyes, and was able to remove from the environment and accumulate in its cells the dyes up to a significant percentage (crystal violet was removed by about 80% and malachite green by 90%).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Magnesiumsulfat, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Magnesiumsulfat -Lösung, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Magnesiumsulfat -Lösung, BioUltra, Molecular Biology
Sigma-Aldrich
Triphenylmethan, 99%
Sigma-Aldrich
Magnesiumsulfat, ≥99.99% trace metals basis
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis