Merck
  • Elucidation of the defence mechanism in microalgae Chlorella sorokiniana under mercury exposure. Identification of Hg-phytochelatins.
Elucidation of the defence mechanism in microalgae Chlorella sorokiniana under mercury exposure. Identification of Hg-phytochelatins.
Chemico-biological interactions (2015-06-17)
Verónica Gómez-Jacinto, Tamara García-Barrera, José Luis Gómez-Ariza, Inés Garbayo-Nores, Carlos Vílchez-Lobato
ZUSAMMENFASSUNG
Algae and aquatic macrophytes are capable of accumulating heavy metals up to concentrations several orders of magnitude higher than those existing in their surrounding environment. Investigation of mercury toxicology in microalgae is of great interest from ecological point of view, since they could be used as bioindicator to evaluate aquatic ecosystems affected by Hg pollution. In this study, we have performed an exposure experiment focused on the biological response of microalgae Chlorella sorokiniana, a unicellular model organism, to Hg-induced toxicity. The culture was exposed to different concentrations of this element for nine days, namely 0.5, 1, 5 and 10mg L(-1) of HgCl2 (as Hg). To achieve a better understanding of the biological mechanisms triggered by Hg-induced toxicity in this alga a metallomic approach based on SEC-ICP-ORS-MS was applied to survey biomarkers of biological response to mercury contamination in surface water. In addition, the combination of RP-HPLC-ICP-ORS-MS and RP-HPLC-ESI-QqQ-TOF-MS was applied to identify, for the first time, two Hg-binding phytochelatins in this aquatic organism, using cell extracts from microalgae exposed to inorganic mercury.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natriumbicarbonat, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Kaliumphosphat, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥98.5% (GC)
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ammoniumacetat, Molecular Biology, ≥98%
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Kaliumphosphat, ReagentPlus®
Sigma-Aldrich
Ammoniumacetat -Lösung, Molecular Biology, 7.5 M
Sigma-Aldrich
Phenylmethansulfonylfluorid, ≥99.0% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Kaliumphosphat, Molecular Biology, ≥98.0%
Sigma-Aldrich
Ammoniak, anhydrous, ≥99.98%
Sigma-Aldrich
Ammoniumacetat, 99.999% trace metals basis
Sigma-Aldrich
Ammoniak -Lösung, 4 M in methanol
Sigma-Aldrich
Kaliumnitrat, BioReagent, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Ammoniak -Lösung, 0.4 M in THF
Sigma-Aldrich
Natriumbicarbonat, BioXtra, 99.5-100.5%
Sigma-Aldrich
Ammoniumacetat, reagent grade, ≥98%
Sigma-Aldrich
Ammoniumacetat, BioXtra, ≥98%
Sigma-Aldrich
Kaliumphosphat, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder
Sigma-Aldrich
Kaliumnitrat, 99.999% trace metals basis
Sigma-Aldrich
Kaliumphosphat, 99.99% trace metals basis
Sigma-Aldrich
Kaliumnitrat, BioXtra, ≥99.0%
Sigma-Aldrich
Kaliumnitrat, 99.99% trace metals basis
Sigma-Aldrich
Natriumbicarbonat-12C, 13C-depleted, 99.9 atom % 12C
Sigma-Aldrich
Ammoniak-14N, 99.99 atom % 14N