Merck
  • One-Dimensional Sliding of p53 Along DNA Is Accelerated in the Presence of Ca(2+) or Mg(2+) at Millimolar Concentrations.
One-Dimensional Sliding of p53 Along DNA Is Accelerated in the Presence of Ca(2+) or Mg(2+) at Millimolar Concentrations.
Journal of molecular biology (2015-07-07)
Agato Murata, Yuji Ito, Risa Kashima, Saori Kanbayashi, Kei Nanatani, Chihiro Igarashi, Masaki Okumura, Kenji Inaba, Takashi Tokino, Satoshi Takahashi, Kiyoto Kamagata
ZUSAMMENFASSUNG
One-dimensional (1D) sliding of the tumor suppressor p53 along DNA is an essential dynamics required for its efficient search for the binding sites in the genome. To address how the search process of p53 is affected by the changes in the concentration of Mg(2+) and Ca(2+) after the cell damages, we investigated its sliding dynamics at different concentrations of the divalent cations. The 1D sliding trajectories of p53 along the stretched DNA were measured by using single-molecule fluorescence microscopy. The averaged diffusion coefficient calculated from the mean square displacement of p53 on DNA increased significantly at the higher concentration of Mg(2+) or Ca(2+), indicating that the divalent cations accelerate the sliding likely by weakening the DNA-p53 interaction. In addition, two distributions were identified in the displacement of the observed trajectories of p53, demonstrating the presence of the fast and slow sliding modes having large and small diffusion coefficients, respectively. A coreless mutant of p53, in which the core domain was deleted, showed only a single mode whose diffusion coefficient is about twice that of the fast mode for the full-length p53. Thus, the two modes are likely the result of the tight and loose interactions between the core domain of p53 and DNA. These results demonstrated clearly that the 1D sliding dynamics of p53 is strongly dependent on the concentration of Mg(2+) and Ca(2+), which maintains the search distance of p53 along DNA in cells that lost homeostatic control of the divalent cations.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
DL-Dithiothreitol -Lösung, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol -Lösung, 1 M in H2O
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylendiamintetraessigsäure, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure -Lösung, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylendiamintetraessigsäure, 99.995% trace metals basis
Sigma-Aldrich
Ethylendiamintetraessigsäure, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylendiamintetraessigsäure, purified grade, ≥98.5%, powder