Merck
  • Modified in vivo comet assay detects the genotoxic potential of 14-hydroxycodeinone, an α,β-unsaturated ketone in oxycodone.
Modified in vivo comet assay detects the genotoxic potential of 14-hydroxycodeinone, an α,β-unsaturated ketone in oxycodone.
Environmental and molecular mutagenesis (2015-04-29)
Kamala Pant, Nicholas Roden, Charles Zhang, Shannon Bruce, Craig Wood, Kimberly Pendino
ZUSAMMENFASSUNG
14-Hydroxycodeinone (14-HC) is an α,β-unsaturated ketone impurity found in oxycodone drug substance and has a structural alert for genotoxicity. 14-HC was tested in a combined Modified and Standard Comet Assay to determine if the slight decrease in % Tail DNA noted in a previously conducted Standard Comet Assay with 14-HC could be magnified to clarify if the response was due to cross-linking activity. One limitation of the Standard Comet Assay is that DNA cross-links cannot be reliably detected. However, under certain modified testing conditions, DNA cross-links and chemical moieties that elicit such cross-links can be elucidated. One such modification involves the induction of additional breakages of DNA strands by gamma or X-ray irradiation. To determine if 14-HC is a DNA crosslinker in vivo, a Modified Comet Assay was conducted using X-ray irradiation as the modification to visualize crosslinking activity. In this assay, 14-HC was administered orally to mice up to 320 mg/kg/day. Results showed a statistically significant reduction in percent tail DNA in duodenal cells at 320 mg/kg/day, with a nonstatistically significant but dose-related reduction in percent tail DNA also observed at the mid dose of 160 mg/kg/day. Similar decreases were not observed in cells from the liver or stomach, and no increases in percent tail DNA were noted for any tissue in the concomitantly conducted Standard Comet Assay. Taken together, 14-HC was identified as a cross-linking agent in the duodenum in the Modified Comet Assay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, ACS reagent, ≥99.9%
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Natriumhydroxid, ACS reagent, ≥97.0%, pellets
Sigma-Aldrich
Natriumhydroxid, reagent grade, ≥98%, pellets (anhydrous)
Sigma-Aldrich
Dimethylsulfoxid, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Natronlauge, 50% in H2O
Sigma-Aldrich
Salzsäure, ACS reagent, 37%
Sigma-Aldrich
Chlorwasserstoff -Lösung, 4.0 M in dioxane
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natronlauge, BioUltra, Molecular Biology, 10 M in H2O
Sigma-Aldrich
Natriumchlorid, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Natronlauge, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Salzsäure -Lösung, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumchlorid -Lösung, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Natriumchlorid, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Natriumchlorid, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Natriumchlorid -Lösung, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Natriumhydroxid, BioXtra, ≥98% (acidimetric), pellets (anhydrous)
Sigma-Aldrich
Natriumhydroxid, puriss., meets analytical specification of Ph. Eur., BP, NF, E524, 98-100.5%, pellets
Sigma-Aldrich
Salzsäure, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Salzsäure, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Chlorwasserstoff -Lösung, 2.0 M in diethyl ether
Sigma-Aldrich
Ethylendiamintetraessigsäure, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Natriumhydroxid, reagent grade, 97%, powder
Sigma-Aldrich
Salzsäure, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Natriumhydroxid, pellets, semiconductor grade, 99.99% trace metals basis