Merck
[18F]fluoromethylcholine as a chemotherapy response read-out in prostate cancer cells.
Molecular imaging and biology : MIB : the official publication of the Academy of Molecular Imaging (2014-11-20)
Daniela E Oprea-Lager, Mitchell P van Kanten, Reindert J A van Moorselaar, Alfons J M van den Eertwegh, Peter M van de Ven, Irene V Bijnsdorp, Otto S Hoekstra, Albert A Geldof
ZUSAMMENFASSUNG
The objective of the present study is to determine whether uptake of [(18)F]fluoromethylcholine ([(18)F]FCH) in comparison with 2-deoxy-2-[(18)F]fluoro-D-glucose ([(18)F]FDG) accurately reflects chemotherapy efficacy at the tumor cell level in prostate cancer (PC). The effects of docetaxel and cabazitaxel on viable tumor cell number were explored in four PC cell lines. Cellular uptake of [(18)F]FDG and [(18)F]FCH was compared with the effects measured using sulforhodamine B (SRB) assay, cell counting and colony formation assay (CFA), as proximators of viable tumor cell number. Agreement between uptake and cell numbers was assessed by Bland-Altman plots. [(18)F]FCH uptake in all PC cell lines significantly correlated to the cell numbers surviving the respective drug concentrations. Bland-Altman analysis showed that [(18)F]FDG uptake resulted in signal overestimation and higher variability after chemotherapy. [(18)F]FCH uptake correlates well with viable tumor cell numbers remaining after docetaxel and cabazitaxel exposure. Radiolabeled choline is a potential response monitoring biomarker after chemotherapy for PC.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Natriumpyruvat, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Natriumpyruvat, ReagentPlus®, ≥99%
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
Sulforhodamin B Mononatriumsalz, Dye content 75 %
Sigma-Aldrich
Natriumpyruvat, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sulforhodamin B Natriumsalz, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Natriumpyruvat, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Sulforhodamin B Natriumsalz, Technical grade
Sigma-Aldrich
Natriumpyruvat, BioXtra, ≥99%
Sigma-Aldrich
Natriumpyruvat, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®