Merck
Reshaping of Human Macrophage Polarization through Modulation of Glucose Catabolic Pathways.
Journal of immunology (Baltimore, Md. : 1950) (2015-07-26)
Elena Izquierdo, Víctor Delgado Cuevas, Salvador Fernández-Arroyo, Marta Riera-Borrull, Emmanuel Orta-Zavalza, Jorge Joven, Eduardo Rial, Angel L Corbi, María M Escribese
ZUSAMMENFASSUNG
Macrophages integrate information from the tissue microenvironment and adjust their effector functions according to the prevalent extracellular stimuli. Therefore, macrophages can acquire a variety of activation (polarization) states, and this functional plasticity allows the adequate initiation, regulation, and resolution of inflammatory responses. Modulation of the glucose metabolism contributes to the macrophage adaptation to the surrounding cytokine milieu, as exemplified by the distinct glucose catabolism of macrophages exposed to LPS/IFN-γ or IL-4. To dissect the acquisition of macrophage effector functions in the absence of activating cytokines, we assessed the bioenergetic profile of macrophages generated in the presence of GM-CSF (GM-MØ) or M-CSF (M-MØ), which do not release pro- or anti-inflammatory cytokines unless subjected to additional activating stimuli. Compared to M-MØ, GM-MØ displayed higher oxygen consumption rate and aerobic glycolysis (extracellular acidification rate [ECAR]), as well as higher expression of genes encoding glycolytic enzymes. However, M-MØ exhibited a significantly higher oxygen consumption rate/ECAR ratio. Surprisingly, whereas aerobic glycolysis positively regulated IL1B, TNF, and INHBA mRNA expression in both macrophage subtypes, mitochondrial respiration negatively affected IL6, IL1B, TNF, and CXCL10 mRNA expression in M-MØ. The physiological significance of these results became evident under low oxygen tensions, as hypoxia enhanced ECAR in M-MØ via HIF-1α and HIF-2α, increased expression of glycolytic enzymes and GM-MØ-specific genes, and diminished expression of M-MØ-associated genes. Therefore, our data indicate that GM-MØ and M-MØ display distinct bioenergetic profiles, and that hypoxia triggers a transcriptomic switch in macrophages by promoting a HIF-1α/HIF-2α-dependent increase in ECAR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dimethylsulfoxid, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethylsulfoxid, Molecular Biology
Sigma-Aldrich
Dimethylsulfoxid, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethylsulfoxid, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethylsulfoxid, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
L-Glutamin, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, 98%
Sigma-Aldrich
Thiazolylblau-Tetrazoliumbromid, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Carbonylcyanid-4-(trifluormethoxy)phenylhydrazon, ≥98% (HPLC), powder
Sigma-Aldrich
Rotenon, ≥95%
Sigma-Aldrich
L-Glutamin, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Dimethylsulfoxid, BioUltra, Molecular Biology, ≥99.5% (GC)
SAFC
L-Glutamin
Sigma-Aldrich
Dimethylsulfoxid, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethylsulfoxid, PCR Reagent
Sigma-Aldrich
L-Glutamin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
1,2,4,5-Tetramethylbenzen, 95%
Sigma-Aldrich
Natrium-Oxamat, ≥98%
Sigma-Aldrich
L-Glutamin, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamin
Sigma-Aldrich
8-Octanoyloxypyren-1,3,6-Trisulfonsäure Trinatriumsalz, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethylsulfoxid, ≥99.6%, ReagentPlus®