Merck
  • Mild guanidinoacetate increase under partial guanidinoacetate methyltransferase deficiency strongly affects brain cell development.
Mild guanidinoacetate increase under partial guanidinoacetate methyltransferase deficiency strongly affects brain cell development.
Neurobiology of disease (2015-04-22)
Layane Hanna-El-Daher, Elidie Béard, Hugues Henry, Liliane Tenenbaum, Olivier Braissant
ZUSAMMENFASSUNG
Among cerebral creatine deficiency syndromes, guanidinoacetate methyltransferase (GAMT) deficiency can present the most severe symptoms, and is characterized by neurocognitive dysfunction due to creatine deficiency and accumulation of guanidinoacetate in the brain. So far, every patient was found with negligible GAMT activity. However, GAMT deficiency is thought under-diagnosed, in particular due to unforeseen mutations allowing sufficient residual activity avoiding creatine deficiency, but enough guanidinoacetate accumulation to be toxic. With poorly known GAA-specific neuropathological mechanisms, we developed an RNAi-induced partial GAMT deficiency in organotypic rat brain cell cultures. As expected, the 85% decrease of GAMT protein was insufficient to cause creatine deficiency, but generated guanidinoacetate accumulation causing axonal hypersprouting and decrease in natural apoptosis, followed by induction of non-apoptotic cell death. Specific guanidinoacetate-induced effects were completely prevented by creatine co-treatment. We show that guanidinoacetate accumulation without creatine deficiency is sufficient to affect CNS development, and suggest that additional partial GAMT deficiencies, which may not show the classical brain creatine deficiency, may be discovered through guanidinoacetate measurement.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Anti-NeuN-Antikörper, Klon A60, clone A60, Chemicon®, from mouse
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
Ameisensäure, ≥95%, FCC, FG
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Anti-β-Tubulin III-Antikörper (neuronal), monoklonaler Antikörper der Maus in Maus hergestellte Antikörper, ~1.0 mg/mL, clone 2G10, purified from hybridoma cell culture
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
Anti-Wachstum-assoziiertes-Protein-43 (GAP-43)-Antikörper, Chemicon®, from rabbit
SAFC
MOPS
Sigma-Aldrich
Anti-Neurofilament-160-kDa-Antikörper, Klon NN18, clone NN18, Chemicon®, from mouse
Sigma-Aldrich
Anti-MAP2A-Antikörper, AP20, ascites fluid, clone AP20, Chemicon®
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%